Skip to Content
Merck
All Photos(1)

Key Documents

SML0642

Sigma-Aldrich

Chaetoglobosin A

from Chaetomium globosum, ≥98% (HPLC)

Synonym(s):

4,7,14,14a,15,15a,16a,16b-Octahydro-7-hydroxy-14-(1H-indol-3-ylmethyl)-4,6,15,15a-tetramethyl-, 3H-Cyclotridec[d]oxireno[f]isoindole-8,11,12(13H)-trione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N2O5
CAS Number:
Molecular Weight:
528.64
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Chaetomium globosum

Quality Level

Assay

≥98% (HPLC)

solubility

DMSO: soluble 10 mg/mL
methanol: soluble 10 mg/mL

storage temp.

−20°C

InChI

1S/C32H36N2O5/c1-17-8-7-10-22-29-31(4,39-29)19(3)27-24(15-20-16-33-23-11-6-5-9-21(20)23)34-30(38)32(22,27)26(36)13-12-25(35)28(37)18(2)14-17/h5-7,9-14,16-17,19,22,24,27-29,33,37H,8,15H2,1-4H3,(H,34,38)/b10-7+,13-12+,18-14+/t17-,19-,22?,24-,27-,28+,29-,31+,32+/m0/s1

InChI key

OUMWCYMRLMEZJH-VIRVUJSBSA-N

Application

Chaetoglobosin A has been used as a standard for the detection and quantification of chaetoglobosin A from Chaetomium globosum by HPLC.

Biochem/physiol Actions

Chaetomium globosum is a fungus commonly found in water-damaged walls and causes Sick Building Syndrome (SBS). Chaetoglobosin A is a mycotoxic cytochalasin. Over 40 chaetoglobosins were characterized, many of which show acute toxicity to mammals, as well as cytotoxicity to human cancer cell lines such as KB, K562, MCF-7, and HepG2.

Cytochalasins were also found to inhibit glucose transport in human erythrocytes by binding to the glucose carrier on the erythrocyte membrane. Chaetoglobosin A was found to inhibit nematode proliferation and egg hatching. It also causes mortality of second stage juveniles of Meloidogyne incognita.

Other Notes

Store the product sealed at −20 °C. Under these conditions the product is stable for at least 2 years.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A PKS gene, pks-1, is involved in chaetoglobosin biosynthesis,
pigmentation and sporulation in Chaetomium globosum
HU Yang
Life Sciences (2012)
Gang Ding et al.
Journal of natural products, 69(2), 302-304 (2006-02-28)
A new cytotoxic cytochalasan-based alkaloid named chaetoglobosin U (1), along with four known analogues, chaetoglobosins C (2), F (3), and E (5) and penochalasin A (4), has been characterized from the EtOAc extract of a solid culture of Chaetomium globosum
Huan Dou et al.
Biological & pharmaceutical bulletin, 34(12), 1864-1873 (2011-12-02)
Chaetoglobosin Fex (Cha Fex), a cytochalasan-based alkaloid, was isolated from marine-derived endophytic fungus Chaetomium globosum QEN-14. The knowledge of its biological function is still limited. We investigated the effects and mechanism of Cha Fex on inflammatory mediators via Toll-like receptor
Santi Thohinung et al.
Archives of pharmacal research, 33(8), 1135-1141 (2010-08-31)
Nine 10-(indol-3-yl)-[13]cytochalasans such as a new chaetoglobosin V (1); two new natural products, prochaetoglobosin III (2) and prochaetoglobosin III(ed) (3); six known chaetoglobosins B-D (4-6), F (7), and G (8) and isochaetoglobosin D (9) in addition to two known sterols
Tatyana S Sidorova et al.
Pharmaceutical research, 25(6), 1297-1308 (2007-11-28)
The purpose of the present study was to identify the biochemical mechanism(s) of the preventative and reversal effects of Chaetoglobosin K (ChK), a bioactive natural product, on inhibition of gap junction-mediated communication and connexin phosphorylation by the tumor promoting organochlorine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service