Skip to Content
Merck
All Photos(1)

Key Documents

SML0521

Sigma-Aldrich

ML 210

≥98% (HPLC)

Synonym(s):

CID 49766530, ML-210, [4-[Bis(4-chlorophenyl)methyl]piperazin-1-yl]-(5-methyl-4-nitro-1,2-oxazol-3-yl)methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20Cl2N4O4
CAS Number:
Molecular Weight:
475.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

2-8°C

SMILES string

Cc1onc(C(=O)N2CCN(CC2)C(c3ccc(Cl)cc3)c4ccc(Cl)cc4)c1[N+]([O-])=O

InChI

1S/C22H20Cl2N4O4/c1-14-20(28(30)31)19(25-32-14)22(29)27-12-10-26(11-13-27)21(15-2-6-17(23)7-3-15)16-4-8-18(24)9-5-16/h2-9,21H,10-13H2,1H3

InChI key

VIBHJPDPEVVDTB-UHFFFAOYSA-N

Application

ML 210 has been used as a glutathione peroxidase 4 (GPX4) inhibitor to induce ferroptosis in cancer cells. It has also been used as a GPX4 inhibitor to examine whether pharmacological inhibition of GPX4 altered prominin2 expression and impacted ferroptosis in adherent MCF10A and Hs578t cells.

Biochem/physiol Actions

ML 210 acts as a selenoenzyme glutathione peroxidase 4 (GPX4) inhibitor. It exhibits cytotoxicity against few ovarian cancer cell lines.
ML 210 induces non-apoptotic cell death in tumor cells expressing the RAS oncogene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amrita Basu et al.
Cell, 154(5), 1151-1161 (2013-09-03)
The high rate of clinical response to protein-kinase-targeting drugs matched to cancer patients with specific genomic alterations has prompted efforts to use cancer cell line (CCL) profiling to identify additional biomarkers of small-molecule sensitivities. We have quantitatively measured the sensitivity
Jianling Bi et al.
Cell death & disease, 10(10), 682-682 (2019-09-19)
Ferroptosis is an iron-dependent, non-apoptotic form of regulated cell death driven by lipid hydroperoxides within biological membranes. Although therapy-resistant mesenchymal-high cancers are particularly vulnerable to ferroptosis inducers, especially phospholipid glutathione peroxidase 4 (GPx4) inhibitors, the underlying mechanism is yet to
Leslie Magtanong et al.
Cell chemical biology, 29(9), 1409-1418 (2022-07-10)
Ferroptosis is an important mediator of pathophysiological cell death and an emerging target for cancer therapy. Whether ferroptosis sensitivity is governed by a single regulatory mechanism is unclear. Here, based on the integration of 24 published chemical genetic screens combined
Yinu Wang et al.
Cancer research, 81(2), 384-399 (2020-11-12)
Defining traits of platinum-tolerant cancer cells could expose new treatment vulnerabilities. Here, new markers associated with platinum-tolerant cells and tumors were identified using in vitro and in vivo ovarian cancer models treated repetitively with carboplatin and validated in human specimens.
Ji-Yoon Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(51), 32433-32442 (2020-12-09)
Ferroptosis is an iron-dependent regulated necrosis mediated by lipid peroxidation. Cancer cells survive under metabolic stress conditions by altering lipid metabolism, which may alter their sensitivity to ferroptosis. However, the association between lipid metabolism and ferroptosis is not completely understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service