Skip to Content
Merck
All Photos(1)

Key Documents

SML0499

Sigma-Aldrich

NCTT-956

≥97% (NMR)

Synonym(s):

N-[(8-Hydroxy-5-nitro-7-quinolinyl)-2-thienylmethyl]-propanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N3O4S
CAS Number:
Molecular Weight:
357.38
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (NMR)

form

powder

color

light orange to dark orange

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

CCC(=O)NC(c1cccs1)c2cc([N+]([O-])=O)c3cccnc3c2O

InChI

1S/C17H15N3O4S/c1-2-14(21)19-15(13-6-4-8-25-13)11-9-12(20(23)24)10-5-3-7-18-16(10)17(11)22/h3-9,15,22H,2H2,1H3,(H,19,21)

InChI key

DEXNORPUTBBNSC-UHFFFAOYSA-N

Biochem/physiol Actions

NCTT-956 is a potent, specific 12-LO inhibitor.
NCTT-956 is a very potent, specific inhibitor of 12-lipoxygenase (12-LO) activity, with an IC50 of 800 nM, no activity against 5-LO and greater than 25-fold selectivity over 15-LO. Specific inhibition of 12-LO by NCTT-956 in platelets blocks PKC activation, aggregation, granule secretion and αIIbB3 activation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luiz Osório Leiria et al.
Cell metabolism, 30(4), 768-783 (2019-07-30)
Distinct oxygenases and their oxylipin products have been shown to participate in thermogenesis by mediating physiological adaptations required to sustain body temperature. Since the role of the lipoxygenase (LOX) family in cold adaptation remains elusive, we aimed to investigate whether
Lisa Mayr et al.
Nature communications, 11(1), 1775-1775 (2020-04-15)
The increased incidence of inflammatory bowel disease (IBD) has become a global phenomenon that could be related to adoption of a Western life-style. Westernization of dietary habits is partly characterized by enrichment with the ω-6 polyunsaturated fatty acid (PUFA) arachidonic
Dennis Y Chuang et al.
Journal of neuroinflammation, 12, 199-199 (2015-11-02)
Oxidative stress and inflammation are important factors contributing to the pathophysiology of numerous neurological disorders, including Alzheimer's disease, Parkinson's disease, acute stroke, and infections of the brain. There is well-established evidence that proinflammatory cytokines and glutamate, as well as reactive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service