Skip to Content
Merck
All Photos(1)

Key Documents

SML0411

Sigma-Aldrich

BAY-X-1005

≥98% (HPLC)

Synonym(s):

α-Cyclopentyl-4-(2-quinolinylmethoxy)-(R)-benzeneacetic acid, (αR)-α-Cyclopentyl-4-(2-quinolinylmethoxy)-benzeneacetic acid, DG 031, Velifapon, Veliflapon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23NO3
CAS Number:
Molecular Weight:
361.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](C1CCCC1)c2ccc(OCc3ccc4ccccc4n3)cc2

InChI

1S/C23H23NO3/c25-23(26)22(17-6-1-2-7-17)18-10-13-20(14-11-18)27-15-19-12-9-16-5-3-4-8-21(16)24-19/h3-5,8-14,17,22H,1-2,6-7,15H2,(H,25,26)/t22-/m1/s1

InChI key

ZEYYDOLCHFETHQ-JOCHJYFZSA-N

Biochem/physiol Actions

Bay-X-1005 is a potent inhibitor of 5-lipoxygenase activating protein (FLAP). Bay-X-1005 inhibits A23187-induced LTB4 production in human leucocytes with an IC50 value of 220 nM, and blocks IgE mediated airway contractions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Titos et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 17(12), 1745-1747 (2003-09-06)
The existence of an increased number of Kupffer cells is recognized as critical in the initiation of the inflammatory cascade leading to liver fibrosis. Because 5-lipoxygenase (5-LO) is a key regulator of cell growth and survival, in the current investigation
P Roger et al.
Mediators of inflammation, 10(1), 33-36 (2001-04-28)
Increased secretion of mucus is a hallmark of many respiratory diseases and contributes significantly to the airflow limitation experienced by many patients. While the current pharmacological approach to reducing mucus and sputum production in patients is limited, clinical studies have
R Müller-Peddinghaus
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 48(4), 529-536 (1998-01-28)
Leukotrienes have been identified in various pathophysiologies. The leukotrienes LTB4 and LTC4 are assigned to inflammation. 5-lipoxygenase inhibitors which inhibit the synthesis of LTA4 being the precursor of both LTB4 and LTC4 appear to have only a limited antiinflammatory potential.
Stefano Fiorucci et al.
British journal of pharmacology, 139(7), 1351-1359 (2003-08-02)
(1) Unlike other nonsteroidal anti-inflammatory drugs that inhibit formation of cyclooxygenase (COX)-dependent eicosanoids, acetylation of COX-2 by aspirin switches eicosanoid biosynthesis from prostaglandin E(2) (PGE(2)) to 15-epi-lipoxin A(4) (15-epi-LXA(4) or aspirin-triggered lipoxin, ATL). ATL formation by activated leukocytes (PMN) requires
Leukotrienes in cardiovascular diseases.
G Folco et al.
American journal of respiratory and critical care medicine, 161(2 Pt 2), S112-S116 (2000-02-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service