Skip to Content
Merck
All Photos(1)

Key Documents

SML0335

Sigma-Aldrich

Remacemide hydrochloride

≥98% (HPLC)

Synonym(s):

2-Amino-N-(1-methyl-1,2-diphenylethyl)-acetamide hydrochloride, FPL 12924AA, PR 934-423A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O · HCl
CAS Number:
Molecular Weight:
304.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: >10 mg/mL

storage temp.

room temp

SMILES string

Cl.CC(Cc1ccccc1)(NC(=O)CN)c2ccccc2

InChI

1S/C17H20N2O.ClH/c1-17(19-16(20)13-18,15-10-6-3-7-11-15)12-14-8-4-2-5-9-14;/h2-11H,12-13,18H2,1H3,(H,19,20);1H

InChI key

HYQMIUSWZXGTCC-UHFFFAOYSA-N

Biochem/physiol Actions

Remacemide HCl is a low affinity NMDA antagonist with anticonvulsant properties. Remacemide also been shown to block voltage-dependent sodium channels.
Remacemide possesses neuroprotective and anti-epileptic actions. It supports reducing the frequency of seizures. Remacemide is also known to be a potential therapeutic for Huntington′s disease.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K A Wesnes et al.
Epilepsy & behavior : E&B, 14(3), 522-528 (2008-12-30)
An international trial comparing remacemide hydrochloride with carbamazepine was undertaken in individuals with newly diagnosed epilepsy using a novel double-blind, parallel-group, double triangular sequential design. Patients with two or more partial or generalized tonic-clonic seizures in the previous year were
Astrid Nehlig et al.
Epilepsy research, 52(3), 253-261 (2003-01-22)
The antiepileptic effects of remacemide were assessed in two models of genetically determined generalized epilepsy. The model of non-convulsive epilepsy used was a model of absence seizures, the GAERS (genetic absence epilepsy rats from Strasbourg), and the model of convulsive
In Vivo Systems: Animal Models of Neurodegeneration
Comprehensive Toxicology, 13 (2010)
D W Chadwick et al.
Seizure, 11(2), 114-123 (2002-04-12)
Remacemide hydrochloride is a low-affinity, non-competitive N-methyl-D-aspartic acid (NMDA) receptor channel blocker, under investigation in epilepsy. This double-blind, placebo-controlled, multicentre study assessed the safety and efficacy of remacemide hydrochloride or placebo, as adjunctive therapy, in 252 adult patients with refractory
L K M Wright et al.
Neurotoxicology and teratology, 29(3), 348-359 (2007-02-13)
The effects of chronic exposure to ketamine or remacemide on the acquisition and performance of food-reinforced operant behaviors was assessed in female Sprague-Dawley rats. Ketamine is an anesthetic N-methyl-D-aspartate (NMDA) receptor antagonist, whereas remacemide is an active central nervous system

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service