Skip to Content
Merck
All Photos(1)

Key Documents

SML0321

Sigma-Aldrich

Cyamemazine

≥98% (HPLC)

Synonym(s):

10-[3-(Dimethylamino)-2-methylpropyl]-10H-phenothiazine-2-carbonitrile, 7204 R. E., 7204 RP, Ciamatil, Cianatil, Cyamemazin, R.P. 7204, TH 2602

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N3S
CAS Number:
Molecular Weight:
323.46
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to yellow-green

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

CC(CN(C)C)CN1c2ccccc2Sc3ccc(cc13)C#N

InChI

1S/C19H21N3S/c1-14(12-21(2)3)13-22-16-6-4-5-7-18(16)23-19-9-8-15(11-20)10-17(19)22/h4-10,14H,12-13H2,1-3H3

InChI key

SLFGIOIONGJGRT-UHFFFAOYSA-N

Application

Cyamemazine has been used as an internal standard and antipsychotic drug to spike plasma samples in mass spectrometry studies. It has also been used as an analytical standard in gas chromatography-tandem mass spectroscopic (GC-MS/MS) analysis.

Biochem/physiol Actions

Cyamemazine belongs to the class of phenothiazine drugs. It is known to treat severe depression, schizophrenia and bipolar disorder.
Cyamemazine is a potent antagonist of 5-HT2C and 5-HT3 receptors with antipsychotic and anxiolytic properties.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurmit Singh et al.
Bioorganic & medicinal chemistry letters, 22(6), 2160-2162 (2012-03-01)
A modified Polonovski reaction has been employed to obtain the N-demethylated metabolite of the neuroleptic drug cyamemazine. The synthesis involves N-oxide formation, isolation of the corresponding N-oxide, and a FeSO(4)·7H(2)O mediated Polonovski reaction to afford the desired monodesmethyl cyamemazine. In
Michel Bourin et al.
CNS drug reviews, 10(3), 219-229 (2004-10-20)
Several studies have suggested that the antipsychotic compound, cyamemazine, possesses anxiolytic properties in humans. The original pharmacological profile of cyamemazine (D(2), 5-HT(2A), 5-HT(2C), and 5-HT(3) receptor antagonist), which was established by binding, microdialysis and behavioral studies, is consistent with these
Sébastien Duband et al.
Forensic science international, 214(1-3), e26-e29 (2011-08-16)
Forensic pathologists are sometimes confronted with microscopic foreign bodies mixed in with soft tissues surrounding wounds and which are thus difficult to identify. This identification, however, could be primordial in investigating a crime and in determining the weapon used. A
Paula Proença et al.
Journal of analytical toxicology, 44(8), 915-922 (2020-08-12)
Antipsychotic drugs (AP) are widely prescribed for the treatment of schizophrenia and psychosis. The pharmacological treatment of schizophrenia is often performed with the simultaneous use of two or more antipsychotic agents to achieve the desired control of psychotic symptoms Available
William Crumb et al.
European journal of pharmacology, 532(3), 270-278 (2006-02-24)
The antipsychotic and anxiolytic phenothiazine, cyamemazine, was investigated for its effects on the hERG (human ether-à-go-go related gene) channel expressed in HEK 293 cells and on native INa, ICa, Ito, Isus, or IK1 of human atrial myocytes. Moreover, cyamemazine and

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service