Skip to Content
Merck
All Photos(1)

Key Documents

SML0141

Sigma-Aldrich

Trilostane

≥98% (HPLC)

Synonym(s):

(4α,5α,17β)-3,17-dihydroxy-4,5-epoxyandrost-2-ene-2-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥17 mg/mL

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CC[C@@]45O[C@@H]4C(O)=C(C[C@]35C)C#N)[C@@H]1CC[C@@H]2O

InChI

1S/C20H27NO3/c1-18-7-6-14-12(13(18)3-4-15(18)22)5-8-20-17(24-20)16(23)11(10-21)9-19(14,20)2/h12-15,17,22-23H,3-9H2,1-2H3/t12-,13-,14-,15-,17+,18-,19+,20+/m0/s1

InChI key

KVJXBPDAXMEYOA-CXANFOAXSA-N

Gene Information

human ... HSD3B2(3284)

General description

Trilostane inhibits the production of adrenal steroids, such as cortisol and aldosterone. It is used to treat aldosteronism.

Application

Trilostane has been used to evaluate its capability to regulate the sex-dependent differences in the lipopolysaccharide (LPS)-induced inflammatory responses of astrocytes.
Trilostane was used to inhibit the activity of 3β-hydroxysteroid dehydrogenase in mouse ovary.

Biochem/physiol Actions

Trilostane is an inhibitor of 3 β-hydroxysteroid dehydrogenase (3-β-HSD or delta 5-delta 4-isomerase), an essential enzyme for the biosynthesis of all classes of hormonal steroids. It has been used in the treatment of Cushing′s syndrome for stopping the production of cortisol, and is currently approved for dogs in the US, but is still a human drug in the UK and other countries. It is being investigated as a possible treatment for both breast cancer and prostate cancer to prevent the synthesis of estrogens and androgens from endogenous precursors. It has also been used to inhibit endogenous production of progesterone in research studies.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nyssa J Reine
Clinical techniques in small animal practice, 22(1), 18-25 (2007-06-05)
Pituitary-dependent hyperadrenocorticism is a common endocrine disorder in dogs in the United States. Once a diagnosis is established, a decision must be made whether or not to pursue treatment, and if so, which medication to use. Historically, mitotane (Lysodren, o,p'-DDD
Chiemi Miura et al.
Reproduction (Cambridge, England), 142(6), 869-877 (2011-10-07)
Gh plays important roles in development, somatic growth and gametogenesis in vertebrates. To determine the physiological role of Gh in reproduction in male teleosts, the expression of genes encoding Gh and the two Gh receptors (Ghrs) during spermatogenesis, and the
Sex-Mediated Differences in LPS Induced Alterations of TNFalpha, IL-10 Expression, and Prostaglandin Synthesis in Primary Astrocytes
Chistyakov D ,et al.
International Journal of Molecular Sciences, 19(9), 2793-2793 (2018)
Emily Cross et al.
Topics in companion animal medicine, 27(1), 8-20 (2012-09-11)
A 12-year-old female spayed domestic short-haired cat presented for lethargy, poor hair coat, alopecia, difficulty walking, and mild polyuria/polydipsia. The cat's skin tore easily in the neck area during routine restraint for blood draw. Physical examination, blood analysis, and ultrasound
J de Gier et al.
Theriogenology, 75(7), 1271-1279 (2011-02-08)
Interference with the pregnancy-maintaining influence of progesterone is the basis of most methods for termination of unwanted pregnancy in dogs. The currently available methods are based on induction of luteolysis or blocking of the progesterone receptor. Inhibition of progesterone synthesis

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service