Skip to Content
Merck
All Photos(1)

Key Documents

SML0117

Sigma-Aldrich

Fosphenytoin disodium salt hydrate

≥98% (HPLC)

Synonym(s):

5,5-Diphenyl-3-[(phosphonooxy)methyl]-2,4-imidazolidinedione disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13N2Na2O6P · xH2O
CAS Number:
Molecular Weight:
406.24 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].[O-]P([O-])(=O)OCN1C(=O)NC(C1=O)(c2ccccc2)c3ccccc3

InChI

1S/C16H15N2O6P.2Na.H2O/c19-14-16(12-7-3-1-4-8-12,13-9-5-2-6-10-13)17-15(20)18(14)11-24-25(21,22)23;;;/h1-10H,11H2,(H,17,20)(H2,21,22,23);;;1H2/q;2*+1;/p-2

InChI key

WPAVXKKGWCHUAK-UHFFFAOYSA-L

Biochem/physiol Actions

Fosphenytoin is a water soluble phenytoin prodrug.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advisers recommend improved labeling for phenytoin, fosphenytoin.
Kate Traynor
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 67(24), 2086-2086 (2010-12-01)
Case 3-2009: a 9-month-old boy with seizures.
Ajit Singh Kashyap et al.
The New England journal of medicine, 360(15), 1572-1572 (2009-04-10)
Clinical pharmacology of parenteral use of antiepileptic drugs.
Meir Bialer
Epilepsia, 48 Suppl 8, 46-48 (2008-03-11)
Linda A Steinbaugh et al.
Epilepsy & behavior : E&B, 23(3), 280-284 (2012-02-22)
Continuous electroencephalography (cEEG) is increasingly used to detect both clinical and subclinical seizures in patients with traumatic brain injury (TBI) or subarachnoid hemorrhage (SAH). We assess whether EEG findings predict outcomes in TBI/SAH patients enrolled in a levetiracetam (LEV) vs.
Effect of an educational initiative on physicians' use of weight-based fosphenytoin loading doses in the emergency department.
April D Miller et al.
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 67(1), 15-16 (2010-01-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service