Skip to Content
Merck
All Photos(1)

Key Documents

SML0070

Sigma-Aldrich

Salsalate

≥98% (HPLC)

Synonym(s):

2-(2-Hydroxybenzoyl)oxybenzoic acid, 2-Hydroxybenzoic acid 2-carboxyphenyl ester, disalicylic acid, salicyloxysalicylic acid, salicylsalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥15 mg/mL

storage temp.

room temp

SMILES string

OC(=O)c1ccccc1OC(=O)c2ccccc2O

InChI

1S/C14H10O5/c15-11-7-3-1-5-9(11)14(18)19-12-8-4-2-6-10(12)13(16)17/h1-8,15H,(H,16,17)

InChI key

WVYADZUPLLSGPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Salsalate was used to study the effect on palmitate-induced insulin resistance and hepatic steatosis in obese rats.

Biochem/physiol Actions

NSAID; Nonacetylated aspirin analog
Salsalate is a nonsteroidal anti-inflammatory drug (NSAID), a nonacetylated salicylate with no more problems of gastrointestinal bleeding than placebo. It inhibits synthesis and release of prostaglandins through the inactivation of cyclooxygenase-1 (COX-1) and COX-2. Salsalate is currently being investigated as a treatment for Type 2 diabetes with possible use to prevent the disease in people at risk. It reduces blood glucose concentrations in patients with type 2 diabetes, as well as in insulin-resistant patients without diabetes.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Corrilynn O Hileman et al.
AIDS (London, England), 24(12), 1958-1961 (2010-07-09)
In this 13-week, open-label, randomized study of the anti-inflammatory salsalate versus usual care, there were no significant improvements in flow-mediated dilation of the brachial artery, endothelial activation, inflammation or coagulation markers, homeostasis model assessment of insulin resistance or lipoproteins with
Tae Woo Jung et al.
Biochemical pharmacology, 86(7), 960-969 (2013-08-21)
Fetuin-A was recently identified as a novel hepatokine which is associated with obesity, insulin resistance and non-alcoholic fatty liver disease. Salsalate, a prodrug of salicylate with an anti-inflammatory effect and lower side effect profile, significantly lowers glucose and triglyceride levels
Gary L Pierce et al.
Circulation, 119(9), 1284-1292 (2009-02-25)
We tested the hypothesis that nuclear factor-kappaB (NF-kappaB) activity contributes to vascular endothelial dysfunction with aging and obesity in humans. We conducted a randomized, double-blind, placebo-controlled crossover study in 14 nondiabetic overweight or obese (body mass index > or =25
J Koska et al.
Diabetologia, 52(3), 385-393 (2008-12-24)
Low-grade inflammation may contribute to obesity-related insulin resistance and has been associated with increased risk of type 2 diabetes mellitus. The present study evaluated whether treatment with salsalate, a traditional anti-inflammatory medication, would improve insulin action in obese non-diabetic individuals.
R F Carvalhal et al.
Biosensors & bioelectronics, 25(10), 2200-2204 (2010-04-07)
The use of an amperometric biosensor for rapid salicylate determination in blood is described. Photolitography was used to make gold electrodes on a polyester film. The plastic microcell was characterized using cyclic voltammetry to demonstrate the electrochemical performance of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service