Skip to Content
Merck
All Photos(1)

Key Documents

SML0032

Sigma-Aldrich

Triacetyl resveratrol

≥98% (HPLC)

Synonym(s):

3,5,4′-Tri-O-acetylresveratrol, 5-[(1E)-2-[4-(acetyloxy)phenyl]ethenyl]-1,3-Benzenediol-1,3-diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O6
CAS Number:
Molecular Weight:
354.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to tan

solubility

DMSO: ≥18 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

room temp

SMILES string

CC(=O)Oc1ccc(cc1)\C=C\c2cc(OC(C)=O)cc(OC(C)=O)c2

InChI

1S/C20H18O6/c1-13(21)24-18-8-6-16(7-9-18)4-5-17-10-19(25-14(2)22)12-20(11-17)26-15(3)23/h4-12H,1-3H3/b5-4+

InChI key

PDAYUJSOJIMKIS-SNAWJCMRSA-N

General description

Resveratrol (RSV) is a natural polyphenolic antioxidant.

Biochem/physiol Actions

Resveratrol prodrug. SIRT inhibitor
Resveratrol prodrug. SIRT inhibitor
Resveratrol (RSV) plays a protective role on the nervous and cardiovascular systems. It is considered as a modulator of energetic metabolism as it controls glucose homeostasis in obese mice, improves fatty acid oxidation (FAO) and promotes the expression of genes that participate in mitochondrial biogenesis.
Triacetyl resveratrol displays superior bioavailability to the parent compound, resveratrol. The compound induces p53 activity and inhibits proliferation in breast and prostate tumor cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resveratrol induces a mitochondrial complex I dependent increase in NADH oxidation responsible for sirtuin activation in liver cells.
Desquiret-Dumas V, et al.
The Journal of Biological Chemistry, 2013 (2013)
Liqiong Ding et al.
Biochemical pharmacology, 171, 113691-113691 (2019-11-11)
Resveratrol is a natural compound found in many plant species that has broad therapeutic benefits. Here, we investigated the effects of resveratrol on the replication of HSV-2. We found that resveratrol accelerated replication of HSV-2 and increased release of progeny
Tammy R Dugas et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 107(3), 646-651 (2018-08-10)
Peripheral artery disease is a cardiovascular disease characterized by a narrowing of arteries that supply blood to the extremities, particularly, the legs. When surgical intervention is warranted, the primary approach is balloon angioplasty. Drug coated balloons (DCB) designed to release
Hyeanjeong Jeong et al.
International journal of molecular sciences, 20(17) (2019-09-05)
Epigenetic remodeling via histone acetylation has become a popular therapeutic strategy to treat Alzheimer's disease (AD). In particular, histone deacetylase (HDAC) inhibitors including M344 and SAHA have been elucidated to be new drug candidates for AD, improving cognitive abilities impaired
Yoojin Lee et al.
Journal of innate immunity, 13(2), 94-106 (2020-09-17)
Natural killer (NK) cells are innate immune lymphocytes that play an important role in anti-viral and anti-tumour immune responses. Several cancer immunotherapy approaches targeting NK cells are currently in clinical or preclinical development. Here, we aimed to find food nutrients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service