Skip to Content
Merck
All Photos(1)

Key Documents

SML0020

Sigma-Aldrich

BX471

≥98% (HPLC)

Synonym(s):

N-[5-chloro-2-[2-[(2R)-4-[(4-fluorophenyl)methyl]-2-methyl-1-piperazinyl]-2-oxoethoxy]phenyl]-urea, ZK-811752

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24ClFN4O3
CAS Number:
Molecular Weight:
434.89
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥25 mg/mL

originator

Bayer

storage temp.

2-8°C

InChI

1S/C21H24ClFN4O3/c1-14-11-26(12-15-2-5-17(23)6-3-15)8-9-27(14)20(28)13-30-19-7-4-16(22)10-18(19)25-21(24)29/h2-7,10,14H,8-9,11-13H2,1H3,(H3,24,25,29)/t14-/m1/s1

InChI key

XQYASZNUFDVMFH-CQSZACIVSA-N

Application

BX471 has been used in chemotaxis assay to study the responses of HEK293 cells expressing human C-C motif chemokine receptor 1 (CCR1) to CC motif ligand 3 (CCL3)-induced cell migration. It has also been used as a CCR1 inhibitor to assess the effects of CCR1 on the migration and epithelial-mesenchymal transition (EMT).

Biochem/physiol Actions

BX471 blocks CCR1 and downregulates the mRNA expression of ICAM-1, P-selectin and E-selectin. It decreases the inflammatory responses in sepsis, prevents monocyte recruitment in inflammation sites in rheumatoid arthritis patients and inhibits interstitial leukocyte recruitment and fibrosis in mouse model of lupus nephritis.
BX471 is an orally active, CCR-1 specific antagonist. It displaces the endogenous CCR-1 ligands MIP-1a, RANTES and MCP-3 (Ki range 1-5 nM). BX471 inhibits CCR-1 effects in leukocytes, including calcium mobilization and migration.

Features and Benefits

This compound is featured on the Chemokine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human MSCs promotes colorectal cancer epithelial-mesenchymal transition and progression via CCL5beta-catenin Slug pathway
Chen K, et al.
Cell Death & Disease, 8(5), e2819-e2819 (2017)
The role of CCR1 and therapeutic effects of anti-CCL3 antibody in herpes simplex virus-induced Behccet's disease mouse model
Sayeed HM, et al.
Immunology (2019)
Min He et al.
American journal of physiology. Gastrointestinal and liver physiology, 292(4), G1173-G1180 (2007-01-20)
Sepsis is a complex clinical syndrome resulting from a harmful host inflammatory response to infection. Chemokines and their receptors play a key role in the pathogenesis of sepsis. BX471 is a potent nonpeptide CC chemokine receptor-1 (CCR1) antagonist in both
Nagarajan Vaidehi et al.
The Journal of biological chemistry, 281(37), 27613-27620 (2006-07-14)
A major challenge in the application of structure-based drug design methods to proteins belonging to the superfamily of G protein-coupled receptors (GPCRs) is the paucity of structural information (1). The 19 chemokine receptors, belonging to the Class A family of
Hans-Joachim Anders et al.
Journal of the American Society of Nephrology : JASN, 15(6), 1504-1513 (2004-05-22)
Slowly progressive renal injury is the major cause for ESRD. The model of progressive immune complex glomerulonephritis in autoimmune MRL(lpr/lpr) mice was used to evaluate whether chemokine receptor CCR1 blockade late in the disease course can affect progression to renal

Articles

We offer many products related to chemokine receptors for your research needs.

We offer many products related to chemokine receptors for your research needs.

We offer many products related to chemokine receptors for your research needs.

We offer many products related to chemokine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service