Skip to Content
Merck
All Photos(2)

Key Documents

SMB00370

Sigma-Aldrich

Tetrahydrocurcumin

≥96% (HPLC)

Synonym(s):

Tetrahydrocurcumin, 1,7-bis(4-hydroxy-3-methoxyphenyl)-3,5-heptanedione, HZIV 81-2, NSC687845, Tetrahydrodiferuloylmethane, 1,7-Bis(4-hydroxy-3-methoxyphenyl)-3,5-heptanedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O6
CAS Number:
Molecular Weight:
372.41
Beilstein:
3485469
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥96% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc(CCC(=O)CC(=O)CCc2ccc(O)c(OC)c2)ccc1O

InChI

1S/C21H24O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h5-6,9-12,24-25H,3-4,7-8,13H2,1-2H3

InChI key

LBTVHXHERHESKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrocurcumin (THC) is a colorless metabolite of curcumin. It has a similar structure to that of curcumin. THC lacks the α, β-unsaturated carbonyl moiety in its chemical structure. It is naturally sourced from the roots of Curcuma zedoaria, Zingiber mioga, and Zingiber officinale.

Application

Tetrahydrocurcumin (THC) has been used as a neuroprotective agent to study its therapeutic potential in improving mitochondrial dysfunction in the ischemic stroke mice model.

Biochem/physiol Actions

Tetrahydrocurcumin is well-studied in treating cancer for its excellent anti-cancer, anti-angiogenic, and chemopreventative activities. It also exerts anti-inflammatory properties. Tetrahydrocurcumin has been shown to have antioxidant and protective effects against diabetes, and vascular dysfunction via alleviation of oxidative stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

385.2 °F

Flash Point(C)

196.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pidaran Murugan et al.
Life sciences, 79(18), 1720-1728 (2006-06-30)
Oxidative stress has been suggested to be a contributory factor in development and complication of diabetes. In the present study, we have investigated the effect of tetrahydrocurcumin (THC), one of the active metabolites of curcumin on antioxidants status in streptozotocin-nicotinamide
Bharat B Aggarwal et al.
Molecules (Basel, Switzerland), 20(1), 185-205 (2014-12-31)
Curcumin (diferuloylmethane), a golden pigment from turmeric, has been linked with antioxidant, anti-inflammatory, anticancer, antiviral, antibacterial, and antidiabetic properties. Most of the these activities have been assigned to methoxy, hydroxyl, α,β-unsaturated carbonyl moiety or to diketone groups present in curcumin.
Sunmin Park et al.
Genes & nutrition, 11, 2-2 (2016-08-03)
Menopausal symptoms are associated with inflammation. Curcumin is a well-known anti-inflammatory bioactive compound from turmeric whereas tetrahydrocurcumin (THC) is a major metabolite of curcumin that may have different efficacies. However, they have not been studied for anti-menopausal symptoms and anti-osteoarthritis
Nandan K Mondal et al.
Neurochemistry international, 122, 120-138 (2018-11-26)
The objectives of this study are to identify the mechanism of mitochondrial dysfunction during cerebral ischemic/reperfusion (I/R) injury and the therapeutic potential of tetrahydrocurcumin (THC) to mitigate mitochondrial dysfunction in experimental stroke model. In our study, 8-10 weeks old male
Jia-Ching Wu et al.
Food & function, 5(1), 12-17 (2013-11-14)
Chemoprevention is a relatively new and promising strategy to prevent human degenerative diseases, including cancer, and is defined as the use of natural dietary compounds and/or synthetic substances to block, inhibit, reverse, or retard the progress of human diseases. Tetrahydrocurcumin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service