Skip to Content
Merck
All Photos(1)

Key Documents

SMB00110

Sigma-Aldrich

Plumieride

≥95% (LC/MS-ELSD)

Synonym(s):

Agoniadin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O12
CAS Number:
Molecular Weight:
470.42
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]3[C@@H]1C=C[C@]34OC(=O)C(=C4)[C@H](C)O

InChI

1S/C21H26O12/c1-8(23)10-5-21(33-18(10)28)4-3-9-11(17(27)29-2)7-30-19(13(9)21)32-20-16(26)15(25)14(24)12(6-22)31-20/h3-5,7-9,12-16,19-20,22-26H,6H2,1-2H3/t8-,9+,12+,13+,14+,15-,16+,19-,20-,21+/m0/s1

InChI key

AOPMSFXOYJXDNJ-IRFSQMTFSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastião de O Rebouças et al.
Journal of ethnopharmacology, 147(2), 474-480 (2013-03-27)
Himatanthus articulatus (Apocynaceae) is a plant native to the Amazon, popularly used to treat external ulcers, tumors, inflammations, cancer, syphilis and malaria. To investigate the in vivo genotoxic and mutagenic potential of this plant, using the comet assay and the
Mahabeer P Dobhal et al.
The Journal of organic chemistry, 69(19), 6165-6172 (2004-09-11)
Plumieride was isolated as one of the major components from the biologically active methanolic extract of the bark of Plumeria bicolor (family Apocynaceae). For investigating the effect of substituents on cytotoxic activity it was modified into a series of compounds.
R S Gupta et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 11(2-3), 169-174 (2004-04-09)
Oral feeding of male rats with plumieride (15 mg/rat/day) for the period of 60 days did not cause any significant change in the body weight of treated rats. However, the weights of testes, epididymides, seminal vesicle and ventral prostate were
M S Abdel-Kader et al.
Journal of natural products, 60(12), 1294-1297 (1998-01-15)
Bioassay-guided fractionation of the EtOAc extract of both Allamanda cathartica and Himatanthus fallax (Apocynaceae) using the Sc-7 yeast strain resulted in the isolation of the weakly cytotoxic isoplumericin and plumericin. In addition, the new lignan 7(R)-methoxy-8-epi-matairesional and three known compounds
T N Tiwari et al.
Phytotherapy research : PTR, 16(4), 393-394 (2002-07-12)
Plumieride has been isolated as an active principle of the leaves of Allamanda cathartica. It showed strong fungitoxicity against some dermatophytes causing dermatomycosis to animals and human beings. It exhibited a noncytotoxic nature against a P(388) mouse leukaemia cell line.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service