Skip to Content
Merck
All Photos(1)

Key Documents

SMB00080

Sigma-Aldrich

Swertiamarin

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O10
CAS Number:
Molecular Weight:
374.34
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC[C@@]3(O)[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C16H22O10/c1-2-7-14(24-6-8-13(21)23-4-3-16(7,8)22)26-15-12(20)11(19)10(18)9(5-17)25-15/h2,6-7,9-12,14-15,17-20,22H,1,3-5H2/t7-,9+,10+,11-,12+,14-,15-,16+/m0/s1

InChI key

HEYZWPRKKUGDCR-QBXMEVCASA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Swertiamarin has shown potential as antiatherogenic agent by inhibiting HMG-CoA reductase activity in high cholesterol fed rats. The high cholesterol fed rats treated with Swertiamarin showed prevention of increased serum total cholesterol, triglycerides, LDLs and VLDLs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jaishree Vaijanathappa et al.
Planta medica, 75(1), 12-17 (2008-11-14)
Swertiamarin, a secoiridoid isolated from the ethyl acetate extract of Enicostemma axillare, was evaluated for antiedematogenic activity using carrageenan-, formalin-, and histamine-induced paw edema methods in rats. In the carrageenan-induced method, the percentages of edema inhibition obtained after 5 h
Shengguo Zhao et al.
Biomedical chromatography : BMC, 18(1), 10-15 (2004-02-12)
Micellar electrokinetic electrophoresis was employed to determine two active components, gentiopicroside (GE) and swertiamarin (SW) in one Tibetan preparation medicine named shiweilongdankeli, two Tibetan herbal medicines named Gentiana rhodantha and Gentiana kitag and three other Chinese Gentiana medicines named Gentiana
M B Patel et al.
Pharmaceutical biology, 49(4), 383-391 (2011-03-12)
Enicostemma hyssopifolium Verdoon (Gentianaceae) has been documented for various therapeutic effects in traditional systems of medicine; the hypoglycemic and hypolipidemic activities are also well reported. Bioactivity guided fractionation of methanol extract of E. hyssopifolium to test the hypothesis that E.
V Jaishree et al.
Journal of ethnopharmacology, 130(1), 103-106 (2010-04-28)
The whole plant of Enicostemma axillare Raynal (Family: Gentianaceae) is used in variety of diseases in traditional Indian system of medicine including hepatic ailments. Swertiamarin isolated from Enicostemma axillare Raynal was evaluated for antioxidant and hepatoprotective activity. Swertiamarin was isolated
Rakesh D Sonawane et al.
Molecular and cellular biochemistry, 340(1-2), 1-6 (2010-03-17)
Diabetic nephropathy (DN) is one of the foremost causes of renal failure and a primary cause of diabetes mellitus related death. Previously, we have reported that aqueous extract of Enicostemma littorale has potential antidiabetic activity. In the present study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service