Skip to Content
Merck
All Photos(1)

Key Documents

SBVT13

Sigma-Aldrich

SB-ratMrp2-HEK293

MRP2 rat vesicles

Synonym(s):

MRP2, Multidrug resistance protein 2, rat ABCC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.84

recombinant

expressed in HEK 293 cells

form

liquid

concentration

5 mg/mL

color

off-white

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Membrane Preparations for Vesicular Transport Assays (VT) are suitable for general drug-efflux transporter interaction studies. Both substrate and inhibitor interactions can be assessed using vesicles. The success of substrate interaction studies strongly depends on the passive permeability of the compound. High permeability substrates might not be detected. Control Membranes with no-, or significantly lower transporter activity are also available.

Application

In the vesicular transport assay so-called "inside-out" membrane vesicles containing ABC transporters are applied. Incubating substrates of the respective efflux transporter in the presence of the inverted membrane vesicles and ATP will allow measuring accumulation of the substrates into the vesicles. In many cases radiolabeled reporter substrates are used but recently SOLVO developed the new PREDIVEZTM Vesicular Transport Kits that use fluorescent reporter substrates.

The standard vesicular transport assay is an inhibitory assay performed with cold test articles. This assay provides information on any interaction between the ABC transporter and the test article. The transport of the reporter substrate is measured in the presence of the test article (typically in 7 concentrations) and IC50 is defined as the concentration inhibiting the transport of the reporter substrate by 50%.

Should radiolabeled form of the investigated compound or adequate analytical methods (LC/MS, HPLC) be available, the vesicular transport assay may be performed in a direct format without the reporter substrate and may identify substrate nature of the test article. The vesicular transport substrate assay is a low throughput assay. It is suitable for low permeability test compounds as high permeability compounds may escape from the vesicles through the lipid bilayer.

Physical form

Supplied as frozen membrane vesicles, containing 5 mg/ml membrane protein, labeled with volume, catalog number (transporter) and date of production.

Legal Information

Distributed for SOLVO Biotechnology, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Rau et al.
Clinical pharmacology and therapeutics, 80(5), 468-476 (2006-11-23)
The adenosine triphosphate-binding cassette (ABC) class transporter ABCC2 (MRP2 [multidrug resistance related protein 2] or cMOAT [canalicular multispecific organic anion transporter]) is involved in the cellular outward transport and elimination of methotrexate. We hypothesized that common genetic variations may contribute
Anne T Nies et al.
Pflugers Archiv : European journal of physiology, 453(5), 643-659 (2006-07-19)
ABCC2 is a member of the multidrug resistance protein subfamily localized exclusively to the apical membrane domain of polarized cells, such as hepatocytes, renal proximal tubule epithelia, and intestinal epithelia. This localization supports the function of ABCC2 in the terminal
Isabelle Benz-de Bretagne et al.
Journal of biomedicine & biotechnology, 2011, 498757-498757 (2011-05-05)
MRP2 encoded by ABCC2 gene is involved in the secretion of numerous drugs and endogenous substrates. Patients with Dubin-Johnson syndrome due to mutation in ABCC2 gene have elevated urinary coproporphyrin ratio (UCP I/(I + III)). Here we investigated whether this
M G Donner et al.
Hepatology (Baltimore, Md.), 34(2), 351-359 (2001-08-02)
Cholestasis induces down-regulation of multidrug resistance protein 2 (Mrp2, symbol Abcc2), which is localized to the canalicular membrane. Given the overlapping substrate specificities of Mrp2 and multidrug resistance protein 3 (Mrp3, symbol Abcc3), we examined the hypothesis of a different
R Evers et al.
British journal of cancer, 83(3), 375-383 (2000-08-06)
The multidrug resistance proteins MRP1 and MRP2 are members of the same subfamily of ATP-binding cassette transporters. Besides organic molecules conjugated to negatively charged ligands, these proteins also transport cytotoxic drugs for which no negatively charged conjugates are known to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service