Skip to Content
Merck
All Photos(1)

Key Documents

SAE0049

Sigma-Aldrich

L-Lactate Dehydrogenase (LDHA)

from human, recombinant, expressed in E. coli, aqueous solution

Synonym(s):

Lactic Dehydrogenase, recombinant from E. coli, α-HBDH, α-hydroxy butyratede hydrogenase, anaerobic lactate dehydrogenase, (S)-Lactate: NAD+ oxidoreductase, L-Lactate Dehydrogenase, Lactate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

biological source

human

Quality Level

recombinant

expressed in E. coli

form

aqueous solution

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

color

colorless

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... LDHA(3939)

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling
The gene LDHA (L-lactate dehydrogenase A chain) is mapped to human chromosome 11p15. It is a subunit of lactate dehydrogenase.In particular, lactic dehydrogenase A (LDHA) is mainly found in skeletal muscle, and for that reason is known as the M subunit. This recombinant form of LDHA has a C-terminal histidine-tag.

Application

L-Lactate Dehydrogenase (LDHA) has been used in in vitro phosphoglycerate mutase 1 (PGAM1) inhibitors screening assay. It has also been used in a colorimetric assay for determining lactate concentration in conditioned media.

Biochem/physiol Actions

L-lactic dehydrogenase catalyzes the conversion of L-lactate into L-pyruvate while reducing NAD+ to NADH and H+.  L-lactate dehydrogenase A chain (LDHA), an enzyme involved in pyruvate metabolism, LDH is responsible for the conversion of pyruvate to lactate, the final step of glycolysis. It is overexpressed in various cancers. LDHA regulates the microenvironment of developing tumors by the hypoxia-inducible factor (HIF)-signaling pathway. LDHA aids in the NAD+ regeneration during the β-oxidation of fatty acid. LDHA (L-lactate dehydrogenase A chain) is responsible for the conversion of pyruvate to lactate, the final step of glycolysis. It is overexpressed in various cancers. In cancer cells, HIF-1a (hypoxia-inducible factor) induces the expression of LDHA, which helps in maintaining glycolysis in cells.
L-lactic dehydrogenase catalyzes the conversion of L-lactate into L-pyruvate while reducing NAD+ to NADH and H+.

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to L-lactate per min at pH 7.5 at 37 °C.

Physical form

Buffered aqueous solution with Hepes (pH 7.5), NaCl and glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and accurate determination of D- and L-lactate, lactose and galactose by enzymatic reactions coupled to formation of a fluorochromophore: Applications in food quality control
F. Shapiro, N. Silanikove
Food Chemistry, 119, 2-2 (2010)
Comparative transcriptome analysis reveals the potential influencing mechanism of dietary astaxanthin on growth and metabolism in Litopenaeus vannamei
Yichao W, et al.
Aquaculture Research (2020)
LDHA is necessary for the tumorigenicity of esophageal squamous cell carcinoma.
Yao F, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 34(1), 25-31 (2013)
Effect of LDHA Inhibition on TNF-?-Induced Cell Migration in Esophageal Cancers
Forkasiewicz A, et al.
International Journal of Molecular Sciences, 23(24) (2022)
The Proteome of Human Liver Peroxisomes: Identification of Five New Peroxisomal Constituents by a Label-Free Quantitative Proteomics Survey
PLoS ONE, 8(2) (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service