Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600208

Sigma-Aldrich

Anti-GAPDH Antibody

monoclonal, RM114

Synonym(s):

Aging-associated gene 9 protein, Glyceraldehyde 3 phosphate dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-GAPDH antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM114, purified immunoglobulin

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM114, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

all

technique(s)

ChIP: 1:200
ELISA: suitable (assay dependent concentration)
flow cytometry: suitable (assay dependent concentration)
immunocytochemistry: 1:200
immunohistochemistry: 1:200
immunoprecipitation (IP): 1:200
western blot: 1:1000

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)

Related Categories

Specificity

This antibody reacts to GAPDH.

Immunogen

A peptide corresponding to the C-terminus of GAPDH

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wilhelm Stoffel et al.
Molecular metabolism, 54, 101335-101335 (2021-09-17)
An increased ω6/ω3-polyunsaturated fatty acid ratio in the current Western diet is regarded as a critical epigenetic nutritional factor in the pathogenesis of several human lifestyle diseases, metabolic syndrome, cardiovascular disease, the central nervous system and the female and male
Guanghong Zhang et al.
Molecular medicine reports, 22(5), 3635-3644 (2020-09-10)
Long non‑coding RNAs (lncRNAs) and microRNAs (miRs) have been reported to regulate disease progression in numerous types of disease, including retinoblastoma (Rb). Therefore, the present study aimed to investigate the effects of the lncRNA FEZ family zinc finger 1 antisense RNA 1 (FEZF1‑AS1) on Rb
Tereza Hrdinova et al.
International journal of oncology, 58(2), 238-250 (2021-01-26)
Chronic myeloid leukemia (CML) is a malignant hematopoietic disorder distinguished by the presence of a BCR‑ABL1 fused oncogene with constitutive kinase activity. Targeted CML therapy by specific tyrosine kinase inhibitors (TKIs) leads to a marked improvement in the survival of
Vipul K Singh et al.
Frontiers in immunology, 13, 865503-865503 (2022-06-28)
Tuberculosis is a leading cause of death in mankind due to infectious agents, and Mycobacterium tuberculosis (Mtb) infects and survives in macrophages (MФs). Although MФs are a major niche, myeloid-derived suppressor cells (MDSCs) are an alternative site for pathogen persistence.
Xiaosong He et al.
Aging and disease (2024-01-25)
AAV-PHP.eB depends on endothelial cells to highly transduce the central nervous system (CNS) and is widely used for intravenous gene therapy. However, the transduction profile and therapeutic efficiency after endothelial cell injury such as ischemic stroke is largely unknown. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service