Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600038

Sigma-Aldrich

Anti-Gamma H2AX (phospho-Ser139) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM224, purified immunoglobulin

Synonym(s):

H2AX Ser139 p, Histone H2AX (phospho ser139)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM224, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 0.2- 1  μg/mL
immunoblotting: 0.5-2 μg/mL
immunocytochemistry: 0.5-2 μg/mL

isotype

IgG

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer139)

Gene Information

human ... H2AX(3014)

General description

Histone H2AX (H2A histone family member X), a histone variant present in almost all eukaryotes, is coded by H2AFX gene. It is mapped to human chromosome 11q23.2–11q23.3.

Specificity

This antibody reacts to Histone H2A.X only when phosphorylated at serine 139. No cross reactivity with other phosphorylated histones.

Immunogen

Synthetic peptide corresponding to Phospho-Histone H2AX (Ser139)

Biochem/physiol Actions

H2AX (H2A histone family member X) helps to maintain genome stability. It plays an important role in localisation and structuring of the repair focus. H2AX also plays an active role in supplying the active repair proteins. γH2AX participates in DSB repair. This protein serves as a signal and target of phosphorylation in the spreading phase.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roisin M McAvera et al.
Scientific reports, 14(1), 8797-8797 (2024-04-17)
Deletions of chromosome 1p (del(1p)) are a recurrent genomic aberration associated with poor outcome in Multiple myeloma (MM.) TRIM33, an E3 ligase and transcriptional co-repressor, is located within a commonly deleted region at 1p13.2. TRIM33 is reported to play a
Anna Solta et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 29(22), 4644-4659 (2023-09-19)
Acquired chemoresistance is a frequent event in small cell lung cancer (SCLC), one of the deadliest human malignancies. Histone deacetylase inhibitors (HDACi) have been shown to synergize with different chemotherapeutic agents including cisplatin. Accordingly, we aimed to investigate the dual
Dong Luo et al.
ACS applied materials & interfaces, 14(13), 14916-14927 (2022-03-23)
Combined radiotherapy (RT) and chemotherapy are prescribed to patients with advanced prostate cancer (PCa) to increase their survival; however, radiation-related side effects and systematic toxicity caused by chemotherapeutic drugs are unavoidable. To improve the precision and efficacy of concurrent RT
Raphael Ceccaldi et al.
Cell stem cell, 11(1), 36-49 (2012-06-12)
Fanconi anemia (FA) is an inherited DNA repair deficiency syndrome. FA patients undergo progressive bone marrow failure (BMF) during childhood, which frequently requires allogeneic hematopoietic stem cell transplantation. The pathogenesis of this BMF has been elusive to date. Here we
Gulam Mohmad Rather et al.
Cancers, 13(5) (2021-03-04)
We tested the antitumor effects of a modified E2F peptide substituting D-Arg for L-Arg, conjugated to penetratin (PEP) against solid tumor cell lines and the CCRF-leukemia cell line, alone and in combination with pemetrexed or with cisplatin. For in-vivo studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service