SAB5500191
Anti-Vimentin Antibody
rabbit monoclonal, SP20
About This Item
Recommended Products
Product Name
Anti-Vimentin antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP20, tissue culture supernatant
biological source
rabbit
Quality Level
recombinant
expressed in proprietary host
conjugate
unconjugated
antibody form
tissue culture supernatant
antibody product type
primary antibodies
clone
SP20, monoclonal
species reactivity
human (tested)
species reactivity (predicted by homology)
bovine, rat, hamster, frog, chicken, mouse
technique(s)
flow cytometry: 1:100
immunoblotting: 1:100
immunohistochemistry: 1:200
isotype
IgG
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... VIM(7431)
Related Categories
General description
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service