Skip to Content
Merck
All Photos(2)

Key Documents

SAB5500162

Sigma-Aldrich

Anti-PI3K P85 antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP62, affinity isolated antibody

Synonym(s):

Anti-AGM7, Anti-GRB1, Anti-IMD36, Anti-p85, Anti-p85-ALPHA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP62, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

frog, bovine, rat, mouse

technique(s)

flow cytometry: 1:100
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PIK3R1(5295)

Related Categories

General description

Phosphatidylinositol 3-kinase phosphorylates the inositol ring of phosphatidylinositol at the 3-prime position. The enzyme comprises a 110kDa catalytic subunit and a regulatory subunit of either 85, 55, or 50kDa. PI3K p85 is a 85kDa regulatory subunit. Phosphatidylinositol 3-kinase plays an important role in the metabolic actions of insulin, and a mutation in this gene has been associated with insulin resistance.

Immunogen

Synthetic peptide from N-terminus of human PI3K p85.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tengfei Wu et al.
Aging, 14(11), 4888-4896 (2022-06-14)
Diabetic kidney disease (DKD), is one of the most common vascular diseases caused by diabetes, eventually progressing into glomerular sclerosis. Qidantang Granule is a traditional Chinese medicine that is commonly used for DKD. However, there is still no experimental evidence
Hongbin Wang et al.
Molecular medicine reports, 16(3), 3351-3356 (2017-07-18)
Breast cancer has one of the highest incidences in females worldwide. Girdin is a novel actin‑binding protein, that induces cell migration and angiogenesis. However, a systematic study of Girdin function in distinct subtypes of breast cancer has not been reported
Chunya Lu et al.
Journal of cellular and molecular medicine, 22(4), 2478-2487 (2018-02-03)
Lung cancer is the leading cause of death in individuals with malignant disease. Non-small-cell lung cancer (NSCLC) is the most common type of lung cancer, and chemotherapy drugs such as cisplatin are the most widely used treatment for this disease.
Cheng Zhang et al.
Oncology letters, 13(4), 2059-2070 (2017-04-30)
Human bone marrow stromal cells (hBMSCs) may contribute to the growth of tyrosine kinase inhibitor (TKI)-resistant chronic myelogenous leukemia (CML). However, there are certain differences in biology between CML and Philadelphia-positive acute lymphoblastic leukemia (Ph+ ALL). Little is known about
Duojie Zhu et al.
Medical oncology (Northwood, London, England), 40(6), 182-182 (2023-05-19)
An increasing amount of evidence has demonstrated the anticancer activity of triterpenes extracted from traditional medicines. Echinocystic acid (EA), a natural triterpene isolated from Eclipta prostrata (L.) L., has previously been shown to exhibit anticancer activity in HepG2 and HL-60

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service