SAB4700868
Monoclonal Anti-CD45 antibody produced in mouse
clone HI30
Synonym(s):
L-CA, LCA, Leukocyte common antigen, Receptor-type tyrosine-protein phosphatase C, T200
About This Item
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
HI30, monoclonal
form
buffered aqueous solution
species reactivity
nonhuman primates, human
concentration
1 mg/mL
technique(s)
flow cytometry: 2 μg/mL
accession no.
NP_002829.2
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... PTPRC(5788)
Related Categories
General description
The previously assigned protein identifier X6R433 has been merged into P08575. Full details can be found on the UniProt database.
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service