Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700727

Sigma-Aldrich

IgM-purified Isotype Control murine

clone PFR-03, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.42

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

PFR-03, monoclonal

form

buffered aqueous solution

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgM

shipped in

wet ice

storage temp.

2-8°C

General description

This mouse IgM monoclonal antibody (clone PFR-03) reacts with undefined epitope on a plant pathogen.
The specificity of staining by monoclonal antibodies to target antigens should be verified by establishing the amount of non-specific antibody binding. Especially at higher concentration (more than 15 μg/ml) the antibody staining usually has consignable background. To this end a non-reactive immunoglobulin of the same isotype is included as a negative control for each specific monoclonal antibody used in a particular immunoassay. The monoclonal antibody PFR-03, generated against a plant pathogen, does not cross-react with other species, and hence all the background that could be observed when working with this antibody would be a result of general nonspecific interactions between an mouse IgM molecule and the respective sample under the particular conditions.

Immunogen

Phytophthora sp.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service