Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700423

Sigma-Aldrich

Monoclonal Anti-CD326 antibody produced in mouse

clone VU-1D9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-EPCAM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

VU-1D9, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EPCAM(4072)

Related Categories

General description

Epithelial cell adhesion molecule (EpCAM) or cluster of differentiation 326 (CD326) is encoded by the 14 kb GA733-2 gene mapped to human chromosome 2p21. Human EpCAM expression is restricted to epithelia of colon, small intestine, lung, kidney, thymus, liver, pancreas, stomach. It is also expressed in several cancers, progenitor and stem cells. CD326 is a transmembrane glycoprotein characterized with an ectodomain, one transmembrane domain, and a 26 residual cytoplasmic domain.
The mouse monoclonal antibody VU-1D9 recognizes an epitope within EGF-like domain I of CD326 / EpCAM, a marker of epithelial lineages. This antibody strongly stains various normal epithelial cells and carcinomas.

Immunogen

Small cell lung carcinoma cell line H69

Application

Suggested working dilution for immunoblotting is 1-4 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Epithelial cell adhesion molecule (EpCAM, CD326) is a transmembrane glycoprotein, involved in several cellular functions, such as epithelial-specific intercellular cell–adhesion, cell signaling, migration, proliferation and differentiation. EpCAM is a dominant surface antigen mapped on human colon carcinoma. It has potential as a diagnostic marker for several carcinomas, including epithelial ovarian cancer (EOC). EpCAM is considered to be a potential target for cancer treatment. EpCAM with the help of components of wnt signal transduction pathway plays a vital role in stem cell signaling.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chikako Zen et al.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society, 17(8), 943-954 (2011-04-15)
We investigated the phenotype of hepatocellular carcinoma (HCC) in livers removed during transplantation after local ablation therapy by transarterial chemoembolization (TACE). This study involved 80 HCC nodules (40 treated with TACE and 40 not treated with local ablation before transplantation)
Carlo Patriarca et al.
Cancer treatment reviews, 38(1), 68-75 (2011-05-18)
Epithelial cell adhesion molecule (EpCAM, CD326) is a pleiotropic molecule that potentially offers therapeutic applications in cancer treatment. Initially described as a dominant surface antigen on human colon carcinoma, it is a transmembrane glycoprotein mediating epithelial-specific intercellular cell-adhesion. Recent data
Luojun Chen et al.
Scientific reports, 8(1), 1188-1188 (2018-01-21)
Circulating tumor cells (CTCs) provide a new approach for auxiliary diagnosis, therapeutic effect evaluation, and prognosis prediction for cancer patients. The epithelial cell adhesion molecule (EpCAM)-based separation method (CellSearch) showed good clinical use in multiple types of cancer. Nevertheless, some
Na Li et al.
Cancer management and research, 11, 10939-10948 (2020-02-06)
Circulating tumor cell (CTC) detection methods based on epithelial cell adhesion molecule (EpCAM) have low detection rates in epithelial ovarian cancer (EOC). Meanwhile, folate receptor alpha (FRα) has high expression in EOC cells. We explored the feasibility of combining FRα
Graham Carpenter et al.
Cancer cell, 15(3), 165-166 (2009-03-03)
The epithelial-specific cell adhesion molecule (EpCAM) modulates cell adhesion and proliferation. Its overexpression correlates with tumor cell proliferation, and EpCAM is a therapeutic target. In the February issue of Nature Cell Biology, Maetzel et al. demonstrate that proliferative responses to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service