Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700394

Sigma-Aldrich

Monoclonal Anti-CD29 antibody produced in mouse

clone MEM-101A, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ITGB1, Anti-Integrin β1 chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-101A, monoclonal

form

buffered aqueous solution

species reactivity

human, pig, canine

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

General description

Cluster of differentiation 29 (CD29), also known as integrin subunit β 1 (ITGB1), is a mechanosensor, encoded by the gene mapped to human chromosome 10p11.22. It is present in both normal cells and tumor-associated cells. The encoded protein has ubiquitous expression with a molecular mass of 130kDa.
The antibody MEM-101A reacts with CD29 (integrin beta1 chain), a 130 kDa single chain type I glycoprotein expressed as a heterodimer (non-covalently associated with the integrin alpha subunits 1-6). CD29 is broadly expressed on majority of hematopoietic and non-hematopoietic cells (leukocytes, platelets, fibroblasts, endothelial cells, epithelial cells and mast cells).

Immunogen

Raji Burkitt′s lymphoma cell line

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Flow cytometry/Cell sorting (1 paper)

Biochem/physiol Actions

Cluster of differentiation 29 (CD29) acts as an adhesive molecule in the development of de novo focal contact, keratinocyte migration and re-epithelialization of wound tissue. CD29 with/without CD27 induce apoptosis, in association with a multifunctional protein galectin (Gal)-3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

β1 Integrin as a Prognostic and Predictive Marker in Triple-Negative Breast Cancer
Yin HL
International Journal of Molecular Sciences, 17 (2016)
Genomic Aberrations Associated with Erlotinib Resistance in Non-small Cell Lung Cancer Cells
Serizawa M
Anticancer Research, 33, 5223-5233 (2013)
CD29 and CD7 Mediate Galectin-3-Induced Type II T-Cell Apoptosis
Fukumori T
Cancer Research, 63, 8302-8311 (2003)
β1 integrin signaling in asymmetric migration of keratinocytes under mechanical stretch in a co-cultured wound repair model
Lu D
BioMedical Engineering OnLine, 15 (2016)
Qifei Wang et al.
Molecular medicine reports, 13(6), 4636-4642 (2016-04-16)
The aim of the present study was to evaluate the different expression levels of thyroid hormone responsive (THRSP; Spot14)/S14 related, Mig12 (S14R) during bone marrow mesenchymal stem cell (BM-MSC) adipogenesis in adolescent idiopathic scoliosis (AIS) patients. MSCs were retrospectively isolated from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service