Skip to Content
Merck
All Photos(6)

Key Documents

SAB4504690

Sigma-Aldrich

Anti-phospho-Bax (pSer184) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer184)

Gene Information

human ... BAX(581)

General description

Bif-1 (BAX-interacting factor 1), a membrane-curvature inducing protein is located on human chromosome 19q13. Bif-1 is also known as SH3GLB1 (SH3 domain containing GRB2 like, endophilin B1)/ Endophilin B1. It belongs to the membrane curvature-driving endophilin protein family.

Immunogen

The antiserum was produced against synthesized peptide derived from human Bax around the phosphorylation site of Ser184.

Immunogen Range: 143-192

Biochem/physiol Actions

Bif-1 (BAX-interacting factor 1) plays a vital role in apoptosis. BAX gene may act as a negative regulator of autophagy in CRC (Colorectal cancer) development. Lack of Bif-1 affects lipid homeostasis and results in obesity followed by insulin resistance.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The substitution of Proline 168 favors Bax oligomerization and stimulates its interaction with LUVs and mitochondria.
Simonyan L, et al.
Biochimica et Biophysica Acta - Biomembranes, 1859(6), 1144-1155 (2017)
Wnt signal transduction pathway and apoptosis: a review.
Pecina-Slaus N
Cancer Cell International, 10(1), 22-22 (2010)
The BAX gene as a candidate for negative autophagy-related genes regulator on mRNA levels in colorectal cancer.
Gil J, et al.
Medical Oncology (Northwood, London, England), 34(2), 16-16 (2017)
Ying Liu et al.
Scientific reports, 6, 20453-20453 (2016-02-10)
Bif-1 is a membrane-curvature inducing protein that is implicated in the regulation of autophagy and tumorigenesis. Here, we report that Bif-1 plays a critical role in regulating lipid catabolism to control the size of lipid droplets and prevent the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service