Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504536

Sigma-Aldrich

Anti-phospho-Src (pTyr418) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ASV, Anti-SRC1, Anti-THC6, Anti-c-SRC, Anti-p60-Src

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 59 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr418)

Gene Information

human ... SRC(6714)

General description

SRC (proto-oncogene, non-receptor tyrosine kinase) belongs to the Src family kinases (SFKs). It contains an amino-terminal domain, conserved Src-homology domains (SH2 and SH3), a tyrosine kinase domain and the carboxyl-terminus. This gene is located on human chromosome 20q11.

Immunogen

The antiserum was produced against synthesized peptide derived from human Src around the phosphorylation site of Tyr418.

Immunogen Range: 386-435

Application

Anti-phospho-Src (pTyr418) antibody has been used in immunofluorescence.

Biochem/physiol Actions

SRC (proto-oncogene, non-receptor tyrosine kinase) controls early HIV-1 (human immunodeficiency virus 1) infection post-entry. This gene participates in fundamental physiological and pathological processes like carcinogenesis and tumor progression. Src kinase activity helps in the migration of endothelial cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SH3 domain of c-Src governs its dynamics at focal adhesions and the cell membrane.
Machiyama H
FEBS Journal, 282(20), 4034-4055 (2015)
madSTORM: a superresolution technique for large-scale multiplexing at single-molecule accuracy.
Yi J, et al.
Molecular Biology of the Cell, 27(22), 3591-3600 (2016)
c-Src activation promotes nasopharyngeal carcinoma metastasis by inducing the epithelial-mesenchymal transition via PI3K/Akt signaling pathway: a new and promising target for NPC
Ke L, et al.
Oncotarget, 7(19), 28340-28355 (2016)
c-SRC protein tyrosine kinase regulates early HIV-1 infection post-entry
McCarthy SD, et al.
AIDS, 30(6), 849-858 (2016)
Subcellular and Dynamic Coordination between Src Activity and Cell Protrusion in Microenvironment.
Zhuo Y
Scientific Reports, 5, 12963-12963 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service