Skip to Content
Merck
All Photos(7)

Key Documents

SAB4504429

Sigma-Aldrich

Anti-phospho-Histone H3.1 (pSer10) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-H3.1, Anti-H3/c, Anti-H3C1, Anti-H3C10, Anti-H3C11, Anti-H3C12, Anti-H3C2, Anti-H3C4, Anti-H3C6, Anti-H3C8, Anti-H3FC, Anti-HIST1H3C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 15 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer10)

General description

Mammals contain three main classes of histone H3 variants: the replicative histones (H3.1 and H3.2), the replacement histone (H3.3) and the centromeric histone (Cenp-A). H3.1 is also called as HIST1H3E. It is mainly expressed in the S phase. HIST1H3E is located on human chromosome 6p22.

Immunogen

The antiserum was produced against synthesized peptide derived from human Histone H3.1 around the phosphorylation site of Ser10.

Immunogen Range: 1-50

Biochem/physiol Actions

Histone proteins are basic building blocks of chromatin. Mutations in histone H3 results in adult cerebellar high-grade gliomas. It controls protein-protein interactions to induce binding of trans-acting factors that drive chromatin condensation. H3.1 acts as a replication-dependent histone.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario A Miranda et al.
Physiological reports, 8(20), e14573-e14573 (2020-10-29)
Maintenance of functional β-cell mass is critical to preventing diabetes, but the physiological mechanisms that cause β-cell populations to thrive or fail in the context of obesity are unknown. High fat-fed SM/J mice spontaneously transition from hyperglycemic-obese to normoglycemic-obese with
A Comprehensive View of the Epigenetic Landscape Part I: DNA Methylation, Passive and Active DNA Demethylation Pathways and Histone Variants.
Sadakierska-Chudy A, et al.
Neurotoxicity Research, 27(1), 84?97-84?97 (2015)
Navrita Mathiah et al.
EMBO reports, 21(11), e50944-e50944 (2020-10-06)
At gastrulation, a subpopulation of epiblast cells constitutes a transient posteriorly located structure called the primitive streak, where cells that undergo epithelial-mesenchymal transition make up the mesoderm and endoderm lineages. Mouse embryo epiblast cells were labelled ubiquitously or in a
Evangéline Despin-Guitard et al.
Nature communications, 15(1), 7364-7364 (2024-08-31)
During the epithelial-mesenchymal transition driving mouse embryo gastrulation, cells divide more frequently at the primitive streak, and half of those divisions happen away from the apical pole. These observations suggest that non-apical mitoses might play a role in cell delamination.
Selective methylation of histone H3 variant H3. 1 regulates heterochromatin replication.
Jacob Y, et al.
Science, 343(6176), 1249-1253 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service