Skip to Content
Merck
All Photos(6)

Key Documents

SAB4503836

Sigma-Aldrich

Anti-phospho-STAT2 (pTyr690) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IMD44, Anti-ISGF-3, Anti-P113, Anti-PTORCH3, Anti-STAT113

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 97 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr690)

Gene Information

human ... STAT2(6773)

General description

STAT2 (signal transducer and activator of transcription 2) is mapped to human chromosome 12.
Signal transducer and activator of transcription 2 (STAT2) belongs to the STAT family. The gene is located on human chromosome 12q13.3. STAT2 consists of 24 exons and has a molecular weight of 113 kDa. It consists of N-terminal domain (N), coiled-coil domain (CC), DNA-binding domain (DBD), linker domain (L), Src homology 2 domain (SH2), tyrosine phosphorylation site (Y) and transcriptional activation domain (TAD).

Immunogen

The antiserum was produced against synthesized peptide derived from human STAT2 around the phosphorylation site of Tyr690.

Immunogen Range: 656-705

Application

Anti-phospho-STAT2 (pTyr690) antibody produced in rabbit has been used in western blotting, immunofluorescence staining and flow cytometric staining.
Anti-phospho-STAT2 (pTyr690) antibody produced in rabbit has been used:
  • for western blotting
  • for immunofluorescence staining
  • for flow cytometric staining

Biochem/physiol Actions

STAT2 (signal transducer and activator of transcription 2) participates in immunity. It plays a major role in interferon signaling pathways.
Signal transducer and activator of transcription 2 (STAT2) is activated by interferon-1 (IFN-I) and IFN-III. It aids protein-protein interactions and is essential for binding IRF9. STAT2 is required for the nuclear export of STAT2-IFN regulatory factor 9 (IRF9) and IFN-stimulated gene factor 3 (ISGF3). It plays a vital role in interactions with STAT1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A haplotype at STAT2 Introgressed from neanderthals and serves as a candidate of positive selection in Papua New Guinea
Mendez FL, et al.
American Journal of Human Genetics, 91(2), 265-274 (2012)
Haochuan Liu et al.
Journal of cellular physiology, 234(10), 17639-17648 (2019-03-12)
Despite the improvements in fracture healing, about 10% of patients undergo abnormal healing. As a tumor suppressor, upregulation of microRNA (miR)-203 has been observed in osteogenic differentiation. Herein, we aimed to explore the functional role of miR-203 in osteoblasts as
Fuquan Jiang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 114, 108814-108814 (2019-04-07)
Bladder cancer is one of common malignancies worldwide. lncRNAs and miRs are reported to play crucial roles in bladder cancer. We aimed to reveal the roles and mechanisms of lncRNA PEG10 and miR-134 in bladder cancer. Transfection was used to
Wei-Hao Lin et al.
Biochemical and biophysical research communications, 514(3), 639-644 (2019-05-12)
It is reported that the genetic variation of DPF3 is a risk factor of breast cancer through large-scale association research. However, the expression, function and mechanism in breast cancer is unknown. We applied qPCR and western blotting to detect the
IL-7Ralpha glutamylation and activation of transcription factor Sall3 promote group 3 ILC development
Liu B, et al.
Nature Communications, 8(1), 231-231 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service