Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502837

Sigma-Aldrich

Anti-SP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

TSFP1, Transcription factor SP1, transcription factor Sp1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 80 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SP1(6667)

General description

Sp1 protein was the first transcription factor to be cloned and characterized. It was first detected in HeLa cells on the basis of its ability to activate the SV40 early promoter transcription. Analysis of structure and function has revealed that Sp1 can be separated into discrete functional domains. The DNA-binding domain consists of three zinc fingers that specifically bind to the GC-box element. Anti-SP1 Antibody detects endogenous levels of total SP1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human SP1.

Immunogen Range: 706-755

Biochem/physiol Actions

Sp1 protein was shown to recognize and bind selectively to a GC-rich consensus sequence (GC-box: GGGCGG or CACCC) that presents in the promoter of several important cellular genes, including Simian vacuolating virus 40 (SV40) early, human immunodeficiency virus-1 (HIV-1), platelet-derived growth factor subunit B (PDGF-B), Myc, c-Src etc. In addition to transcription, Sp1 function has been linked to cell growth, cancer and Huntington disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Shu et al.
Cell death discovery, 8(1), 266-266 (2022-05-19)
Hepatic fibrosis (HF) is caused by chronic hepatic injury and is characterized by hepatic stellate cells (HSCs) activation. Studies focusing on the function of exosomes derived from macrophages in HF progression are limited. This study aims to identify the roles
ShengXing Zheng et al.
Biochemical pharmacology, 177, 113955-113955 (2020-04-07)
Vitamin D regulates cell proliferation, inhibits cytokines release at sites of inflammation and reduces inflammatory responses. In this study, the aim was to investigate whether exogenous vitamin D attenuates LPS-induced lung injury via modulating epithelial cell proliferation, migration, apoptosis and
Structures of zinc finger domains from transcription factor Sp1. Insights into sequence-specific protein-DNA recognition.
Narayan VA
The Journal of Biological Chemistry, 272(12), 7801-7809 (1997)
Mithramycin is a gene-selective Sp1 inhibitor that identifies a biological intersection between cancer and neurodegeneration.
Sleiman SF
The Journal of Neuroscience, 31(18), 6858-6870 (2011)
Decreased association of the transcription factor Sp1 with genes downregulated in Huntington's disease.
Chen-Plotkin AS
Neurobiology of Disease, 22(2), 233-241 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service