Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502398

Sigma-Aldrich

Anti-JNK1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

JNK-46, JNK1-α-2, MAPK8, MK08, Mitogen-activated protein kinase 8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK8(5599)

Related Categories

General description

Anti-JNK1 Antibody detects endogenous levels of total JNK1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human JNK1/2/3.

Immunogen Range: 151-200

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Chen et al.
BMC cardiovascular disorders, 18(1), 72-72 (2018-04-29)
Smooth muscle cells play an important role in the development of atherosclerosis. SHP2 is known to regulate the proliferation and migration of smooth muscle cells. The purpose of this study was to determine whether the SHP2 inhibitor PHPS1 has a
Chunhua Wang et al.
Molecular medicine reports, 16(1), 922-928 (2017-06-01)
In the present study, the protective effect and the potential underlying mechanism of rutaecarpine (Ru) on renal ischemia reperfusion injury (IRI) in rats were investigated. A renal ischemia reperfusion mouse model was established. Ru at 30, 60 mg/kg administered intraperitoneally prior
Xiuli Zhang et al.
International journal of molecular medicine, 35(6), 1747-1754 (2015-04-16)
Zinc (Zn) as an essential dietary element has been indicated in a number of protein functions in the prevention of numerous types of epithelial-to-mesenchymal transition (EMT)-driven fibrosis in vivo. However, relatively little is known regarding its effect in the EMT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service