Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502234

Sigma-Aldrich

Anti-Cytochrome c antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CYC, CYCS, Cytochrome c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 11 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYCS(54205)

General description

Anti-Cytochrome c antibody detects endogenous levels of total Cytochrome c protein.
The cytochrome c gene is mapped to human chromosome 7p15.3. The encoded protein consists of two thioether present between heme and two cysteine residues included in a CXXCH motif (highly conserved). It is localized to mitochondrial intermembrane space.

Immunogen

The antiserum was produced against synthesized peptide derived from human Cytochrome c.

Immunogen Range: 10-59

Application

Anti-Cytochrome c, N-terminal antibody produced in rabbit has been used in western blot analysis.

Biochem/physiol Actions

Cytochrome c is a water-soluble intermediary of electron transport chain. It acts as an ROS (reactive oxygen species) scavenger, and involved in redox-mediated protein import and apoptosis. Cytochrome c has peroxidase activity, contributing towards intrinsic pathway of apoptosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Biosynthesis of Single Thioether c-Type Cytochromes Provides Insight into Mechanisms Intrinsic to Holocytochrome c Synthase (HCCS).
Babbitt SE
Biochemistry, 56(26), 3337-3346 (2017)
Enhancing the peroxidase activity of cytochrome c by mutation of residue 41: implications for the peroxidase mechanism and cytochrome c release.
Josephs TM
The Biochemical Journal, 458(2), 259-265 (2014)
Eman Mohamed Fath et al.
Forensic toxicology (2024-08-09)
The goal of the current study was to clarify the potential molecular mechanism underlying the protective effects of silymarin (SIL) administration against diazinon-induced subacute nephrotoxicity, with a special emphasis on the role of the Kelch-like-associated protein-1 (Keap1)-nuclear factor erythroid 2-related
Nesrine Ebrahim et al.
International journal of molecular sciences, 23(11) (2022-06-11)
Trastuzumab (Trz) is a humanized monoclonal antibody targeting epidermal growth factor receptor 2 (HER2; ErbB2). The combined administration of Trz and doxorubicin (DOX) has shown potent anti-cancer efficacy; however, this regimen may be accompanied by severe cardiac toxicity. Mesenchymal stem
Effect of a K72A Mutation on the Structure, Stability, Dynamics, and Peroxidase Activity of Human Cytochrome c.
Nold SM
Biochemistry, 56(26), 3358-3368 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service