Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501927

Sigma-Aldrich

Anti-MT-ND1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NADH dehydrogenase subunit 1, NADH-ubiquinone oxidoreductase chain 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MT-ND1(4535)

General description

Anti-MT-ND1 Antibody detects endogenous levels of total MT-ND1 protein.
MT-ND1 (mitochondrially encoded NADH dehydrogenase 1mt-Nd1) codes for NADH dehydrogenase-1.

Immunogen

The antiserum was produced against synthesized peptide derived from human MT-ND1.

Immunogen Range: 176-225

Biochem/physiol Actions

MT-ND1 (mitochondrially encoded NADH dehydrogenase 1mt-Nd1) plays a vital role in the electron transport chain of oxidative phosphorylation (OXPHOS). Mutations in MT-ND1 gene is observed in individuals with LHON (leber′s hereditary optic neuropathy). Mutation in MT-ND1 results in dextrocardia, ventriculoarterial discordance and tricuspid atresia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondrial ND1 variants in 1281 Chinese subjects with Leber's hereditary optic neuropathy.
Ji Y, et al.
Investigative Ophthalmology & Visual Science, 57(6), 2377-2389 (2016)
Determination of a New Mutation in MT?ND 1 Gene of a Patient With Dextrocardia, Ventriculoarterial Discordance, and Tricuspid Atresia.
Hatemi AC, et al.
Artificial Organs, 39(1), 83-84 (2015)
Genetic diversities of MT-ND1 and MT-ND2 genes are associated with high-altitude adaptation in yak.
Shi Y, et al.
Mitochondrial DNA. Part A, DNA mapping, sequencing, and analysis, 29(3), 485-494 (2018)
Engy A Abdel-Rahman et al.
Journal of advanced research, 31, 35-47 (2021-07-02)
Incidents of myocardial infarction and sudden cardiac arrest vary with time of the day, but the mechanism for this effect is not clear. We hypothesized that diurnal changes in the ability of cardiac mitochondria to control calcium homeostasis dictate vulnerability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service