Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501901

Sigma-Aldrich

Anti-MMP-14 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MATRIX METALLOPROTEINASE 14, MEMBRANE-TYPE, MEMBRANE-TYPE MATRIX METALLOPROTEINASE 1, MT1-MMP, matrix metalloproteinase 14 (membrane-inserted)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 65 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP14(4323)

General description

Anti-MMP-14 Antibody detects endogenous levels of total MMP-14 protein. Matrix metalloproteinase-14 (MMP-14) is a collagenase localized to the cell membrane. The gene encoding it is present on human chromosome 14.

Immunogen

The antiserum was produced against synthesized peptide derived from human MMP-14.

Immunogen Range: 471-520

Biochem/physiol Actions

Matrix metalloproteinase-14 (MMP-14) aids in the migration of leukocytes and breaks down collagen. Migration of cells through the extracellular matrix is also mediated by it. MMP-14 has been shown to be associated with tuberculosis and glioma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Smeda et al.
Breast cancer research : BCR, 20(1), 86-86 (2018-08-05)
Mesenchymal transformation of pulmonary endothelial cells contributes to the formation of a metastatic microenvironment, but it is not known whether this precedes or follows early metastasis formation. In the present work, we characterize the development of nitric oxide (NO) deficiency
MT1-MMP silencing by an shRNA-armed glioma-targeted conditionally replicative adenovirus (CRAd) improves its anti-glioma efficacy in vitro and in vivo.
Ulasov I
Cancer Letters (2015)
Matrix metalloproteinases in skin.
Kahari VM, Ssaarialho K
Experimental Dermatology (1997)
Membrane Type 1 Matrix Metalloproteinase Regulates Monocyte Migration and Collagen Destruction in Tuberculosis.
Sathyamoorthy T
Journal of Immunology (2015)
Marta Smeda et al.
Oncotarget, 9(25), 17810-17824 (2018-05-01)
Platelet inhibition has been considered an effective strategy for combating cancer metastasis and compromising disease malignancy although recent clinical data provided evidence that long-term platelet inhibition might increase incidence of cancer deaths in initially cancer-free patients. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service