Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501831

Sigma-Aldrich

Anti-TAU antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MAPT, MTBT1, Microtubule-associated protein tau, Neurofibrillary tangle protein, Paired helical filament-tau

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 78 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)

Related Categories

General description

Anti-TAU antibody detects endogenous levels of total TAU protein.
MAPT (microtubule associated protein tau) is located on human chromosome 17q21.3. It is expressed in neurons but is most prominent in axons.

Immunogen

The antiserum was produced against synthesized peptide derived from human Tau.

Immunogen Range: 266-315

Biochem/physiol Actions

Removal of MAPT results in developmental delay and learning disability. It participates in the pathology of Alzheimer′s disease (AD). It helps in the assembly and maintenance of microtubule structure.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahmoud B Maina et al.
Journal of molecular biology, 434(19), 167785-167785 (2022-08-13)
A characteristic hallmark of Alzheimer's Disease (AD) is the pathological aggregation and deposition of tau into paired helical filaments (PHF) in neurofibrillary tangles (NFTs). Oxidative stress is an early event during AD pathogenesis and is associated with tau-mediated AD pathology.
Youssra K Al-Hilaly et al.
Journal of molecular biology, 430(21), 4119-4131 (2018-08-20)
Alzheimer's disease is a tauopathy characterized by pathological fibrillization of tau protein to form the paired helical filaments (PHFs), which constitute neurofibrillary tangles. The methylthioninium (MT) moiety reverses the proteolytic stability of the PHF core and is in clinical development
Rui Duan et al.
Aging, 12(14), 14819-14829 (2020-07-24)
Previously, we revealed that brain Ang-(1-7) deficiency was involved in the pathogenesis of sporadic Alzheimer's disease (AD). We speculated that restoration of brain Ang-(1-7) levels might have a therapeutic effect against AD. However, the relatively short duration of biological effect
Linkage disequilibrium and association of MAPT H1 in Parkinson disease
Skipper L, et al.
American Journal of Human Genetics, 75(4), 669-677 (2004)
The H1c haplotype at the MAPT locus is associated with Alzheimer's disease
Myers AJ, et al.
Human Molecular Genetics, 14(16), 2399-2404 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service