Skip to Content
Merck
All Photos(3)

Key Documents

SAB4501719

Sigma-Aldrich

Anti-LAMA4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

LAMA4, Laminin subunit α-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 202 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMA4(3910)

General description

Anti-LAMA4 Antibody detects endogenous levels of total LAMA4 protein.
Laminin-A4 (LAMA4) is a protein that is expressed at high levels in vivo in clusters of hypertrophic chondrocytes. It is located on human chromosome 6q21.

Immunogen

The antiserum was produced against synthesized peptide derived from human LAMA4.

Immunogen Range: 481-530

Application

Anti-LAMA4 antibody has been used in western blotting, immunohistochemistry and immunocytochemistry.

Biochem/physiol Actions

Laminin-A4 (LAMA4) decreases the amount of cellular protrusions in cultured human chondrocytes. It participates in migration, proliferation and survival of several cells. LAMA4 induces invasion, migration and angiogenesis of trophoblast cell.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Laminin?A4 inhibition on cluster formation of human osteoarthritic chondrocytes.
Moazedi?Fuerst FC, et al.
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society, 34(3), 419-426 (2016)
N Shan et al.
Placenta, 36(8), 809-820 (2015-06-11)
The laminin α4 subunit (LAMA4) has been shown to promote migration, proliferation, and survival of various cell types. This study investigated LAMA4's role in trophoblast cells during placental development. LAMA4 expression was immunohistochemically assessed in the first trimester and term
Laminin α4 (LAMA4) expression promotes trophoblast cell invasion, migration, and angiogenesis, and is lowered in preeclamptic placentas.
Shan N, et al,
Placenta, 36(8), 809?820-809?820 (2015)
The laminin family.
Aumailley M.
Cell adhesion & migration, 7(1), 48-55 (2013)
Anna Goddi et al.
Endocrinology, 163(11) (2022-09-21)
Laminin-α4 (LAMA4) is an extracellular matrix protein implicated in the regulation of adipocyte differentiation and function. Prior research describes a role for LAMA4 in modulating adipocyte thermogenesis and uncoupling protein-1 (UCP1) expression in white adipose; however, the mechanisms involved are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service