Skip to Content
Merck
All Photos(3)

Key Documents

SAB4501309

Sigma-Aldrich

Anti-GR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GCR, GRL, GRL1, NR3C1, glucocorticoid receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 85 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR3C1(2908)

General description

Glucocorticoid receptor (GR) gene/NR3C1 (nuclear receptor subfamily 3 group C member 1) is located on human chromosome 5q31. It belongs to the steroid receptor superfamily that consists of ligand-activated nuclear transcription factors.

Immunogen

The antiserum was produced against synthesized peptide derived from human GR.

Immunogen Range: 181-230

Application

Anti-GR antibody has been used in western blotting.

Biochem/physiol Actions

The glucocorticoid receptor (GR) gene/(NR3C1) is associated with asthma. It is required for glucose homeostasis, hyperinsulinemia and increased fat deposition. It also regulates cell differentiation, nervous and immune system functions.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fulu Ai et al.
Oncology reports, 43(2), 427-436 (2020-01-03)
Glucocorticoids (GCs) are known potent clinical drugs, however, their mode of action is still complex and debatable. Macrophages are the most important target of GCs and play a key role in tumor immunity in vivo, but their relationship is also controversial. In
Ken Kobayashi et al.
Toxicology in vitro : an international journal published in association with BIBRA, 63, 104741-104741 (2019-11-30)
Nicotine from tobacco smoke is absorbed into the bloodstream and transferred into breast milk in breastfeeding mothers. Smoking causes a decrease in breast milk volume, adverse changes to the milk composition, and a shortened lactation period. Breast milk is produced
Yalan Deng et al.
Nature communications, 12(1), 7041-7041 (2021-12-08)
Despite unprecedented responses of some cancers to immune checkpoint blockade (ICB) therapies, the application of checkpoint inhibitors in pancreatic cancer has been unsuccessful. Glucocorticoids and glucocorticoid receptor (GR) signaling are long thought to suppress immunity by acting on immune cells.
Prolactin and glucocorticoid signaling induces lactation-specific tight junctions concurrent with ?-casein expression in mammary epithelial cells
Kobayashi K, et al.
Biochimica et Biophysica Acta, 1863(8), 2006-2016 (2016)
Multiple promoters exist in the human GR gene, one of which is activated by glucocorticoids
Breslin MB, et al.
Molecular Endocrinology, 15(8), 1381-1395 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service