Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500736

Sigma-Aldrich

Anti-4E-BP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

4EBP1, EIF4EBP1, Eukaryotic translation initiation factor 4E binding protein 1, Insulin-stimulated EIF-4E binding protein PHAS-I, P/OKCL.6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 12 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EIF4EBP1(1978)

Related Categories

General description

Anti-4E-BP1 Antibody detects endogenous levels of total 4E-BP1 protein.
The gene E1F4EBP1 (eukaryotic translation initiation factor 4E binding protein 1) is located on human chromosome 8p11. 4E-BP1 is a multifactor regulated multifunctional protein. It belongs to the translation repressor protein family.

Immunogen

The antiserum was produced against synthesized peptide derived from human 4E-BP1.

Immunogen Range: 30-79

Application

Anti-4E-BP1 antibody has been used in western blotting.

Biochem/physiol Actions

E1F4EBP1 (eukaryotic translation initiation factor 4E binding protein 1) is involved in protein synthesis, cell growth and survival. Phosphorylation of E1F4EBP1 releases it from eIF4E (eukaryotic translation initiation factor 4E), thereby resulting in cap-dependent translation. E1F4EBP1 is also involved in tumorigenesis and the gene is overexpressed in various carcinomas. It also serves as a substrate for mTORC1 (mammalian target of rapamycin complex 1).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of zTOR-associated microRNAs in zebrafish embryo treated with rapamycin
Khor ES, et al.
Life Sciences, 150, 67-75 (2016)
4E-BP1, a multifactor regulated multifunctional protein.
Qin X, et al.
Cell Cycle, 15(6), 781-786 (2016)
MicroRNAs 125a and 125b inhibit ovarian cancer cells through post-transcriptional inactivation of EIF4EBP1.
Lee M, et al.
Oncotarget, 7(8), 8726-8726 (2016)
Hui-Ying Huang et al.
Translational cancer research, 11(5), 1076-1088 (2022-06-17)
New and effective chemotherapy or targeted therapy strategies are needed against laryngeal squamous cell carcinoma (LSCC). We aimed to explore the antitumor effect of dual PI3K/mTOR inhibitor combined with autophagy suppression on LSCC and its underlying mechanism. Hep-2 and AMC-HN-8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service