Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500248

Sigma-Aldrich

Anti-DJ-1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Oncogene DJ1, PARK7, Parkinson disease protein 7, Protein DJ-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 19 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARK7(11315)

Related Categories

General description

Anti-DJ-1 Antibody detects endogenous levels of total DJ-1 protein.
DJ-1 (protein deglycase)/PARK7 (parkinsonism associated deglycase) is an oncogene, made of 189 amino acids. It is present in several human tissues and is located on chromosome 1p36.

Immunogen

The antiserum was produced against synthesized peptide derived from human DJ-1.

Immunogen Range: 21-70

Application

Anti-DJ-1 antibody has been used in western blotting.

Biochem/physiol Actions

In lung cancer patients, DJ-1 (protein deglycase) acts as a biomarker for the initial prognosis. It also acts as a serum biomarker for identifying high-risk endometrial cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DJ-1 as a potential biomarker for the early diagnosis in lung cancer patients
Han B, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(6) (2017)
Park7, a novel locus for autosomal recessive early-onset parkinsonism, on chromosome 1p36
van Duijn CM, et al.
American Journal of Human Genetics, 69(3), 629-634 (2001)
DJ-1 is a reliable serum biomarker for discriminating high-risk endometrial cancer
Di Cello A, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(6) (2017)
Lin Cheng et al.
Experimental and therapeutic medicine, 21(6), 557-557 (2021-04-15)
Studies have confirmed that DJ-1 is associated with diseases associated with the nervous system, including Alzheimer's disease (AD). However, the role of DJ-1 in the pathogenesis of AD has not been clarified. To investigate the effect of DJ-1 on brain
Kuldip D Dave et al.
Neurobiology of disease, 70, 190-203 (2014-06-28)
Recessively inherited loss-of-function mutations in the PTEN-induced putative kinase 1(Pink1), DJ-1 (Park7) and Parkin (Park2) genes are linked to familial cases of early-onset Parkinson's disease (PD). As part of its strategy to provide more tools for the research community, The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service