Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500043

Sigma-Aldrich

Anti-G3BP-1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ATP-dependent DNA helicase VIII, G3BP-1, GAP SH3 domain-binding protein 1, HDH-VIII, Ras GTPase-activating protein-binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 52 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... G3BP1(10146)

General description

Anti-G3BP-1 Antibody detects endogenous levels of total G3BP-1 protein.
G3BP1 (ras-GTPase-activating protein SH3 domain-binding protein 1) is a constituent of SGs (stress granules). It is located on human chromosome 5q14.2-5q33.3.

Immunogen

The antiserum was produced against synthesized peptide derived from human G3BP-1.

Immunogen Range: 199-248

Biochem/physiol Actions

G3BP1 (ras-GTPase-activating protein SH3 domain-binding protein 1) blocks the replication of HIV-1 (human immunodeficiency virus) in macrophages and T-cells. In gastric cancer patients, overexpression of G3BP1 leads to imperfect clinical predictions. It is required for the interactions of SG–PB (stress granule-processing bodies).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Role of Ras-GTPase Activating Protein SH3 Domain-Binding Protein G3BP in Adhesion and Migration of 32D Myeloid Progenitor Cells
Schwarz K, et al.
The Open Hematology Journal, 6 (2012)
G3BP1 promotes stress-induced RNA granule interactions to preserve polyadenylated mRNA.
Aulas A, et al.
The Journal of Cell Biology, 209(1), 73-84 (2015)
G3BP1 restricts HIV-1 replication in macrophages and T-cells by sequestering viral RNA
Jimenez VC, et al.
Virology, 486, 94-104 (2015)
Overexpression of Ras-GTPase-activating protein SH3 domain-binding protein 1 correlates with poor prognosis in gastric cancer patients
Min L, et al.
Histopathology, 67(5), 677-688 (2015)
Hideaki Matsuki et al.
Genes to cells : devoted to molecular & cellular mechanisms, 18(2), 135-146 (2013-01-03)
Upon exposure to various environmental stresses such as arsenite, hypoxia, and heat shock, cells inhibit their translation and apoptosis and then repair stress-induced alterations, such as DNA damage and the accumulation of misfolded proteins. These types of stresses induce the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service