Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300468

Sigma-Aldrich

Anti-PTK2B (Ab-402) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CADTK antibody produced in rabbit, Anti-CAKB antibody produced in rabbit, Anti-FADK2 antibody produced in rabbit, Anti-FAK2 antibody produced in rabbit, Anti-PTK2B protein tyrosine kinase 2 beta antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~116 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(D-I-Y-A-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTK2B(2185)

General description

PTK2B/ Pyk2 (proline-rich tyrosine kinase 2β) is a non receptor protein kinase that belongs to the focal adhesion kinase family. It is also known as FAK2 (focal adhesion kinase 2), RAFTK (related adhesion focal tyrosine kinase) and CAKB (cell adhesion kinase beta). Its expression is confined to the endothelium, central nervous system and hematopoietic lineages. It is located on human chromosome 8p21.1.

Immunogen

Peptide sequence around aa. 400-404 (D-I-Y-A-E), according to the protein PTK2B.

Application

Anti-PTK2B (Ab-402) antibody has been used in immunohistofluorescence.

Biochem/physiol Actions

Protein tyrosine kinase 2β(PTK2B) participates in the pathogenesis of Alzheimer′s disease (AD). It induces tumor progression in multiple myeloma. It helps in the multiplication, movement and survival of cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Involved in calcium induced regulation of ion channel and activation of the map kinase signaling pathway. May represent an important signaling intermediate between neuropeptide activated receptors or neurotransmitters that increase calcium flux and the downstream signals that regulate neuronal activity. Interacts with the SH2 domain of Grb2. May phosphorylate the voltage-gated potassium channel protein Kv1.2. Its activation is highly correlated with the stimulation of c-Jun N-terminal kinase activity. Involved in osmotic stress-dependent SNCA 'Tyr-125' phosphorylation.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional screening of Alzheimer risk loci identifies PTK2B as an in vivo modulator and early marker of Tau pathology
Dourlen P, et al.
Molecular Psychiatry, 22(6), 874-883 (2017)
Whole-genome association studies of alcoholism with loci linked to schizophrenia susceptibility
Namkung J, et al.
BMC Genetics, 6 (2005)
A Dual Role for the Nonreceptor Tyrosine Kinase Pyk2 during the Intracellular Trafficking of Human Papillomavirus 16
Gottschalk EY and Meneses PI
Journal of Virology, 89(17), 9103-9114 (2015)
Common variant in PTK2B is associated with late-onset Alzheimer's disease: A replication study and meta-analyses
Li YQ, et al.
Neuroscience Letters, 621, 83-87 (2016)
P Dourlen et al.
Molecular psychiatry, 22(6), 874-883 (2016-04-27)
A recent genome-wide association meta-analysis for Alzheimer's disease (AD) identified 19 risk loci (in addition to APOE) in which the functional genes are unknown. Using Drosophila, we screened 296 constructs targeting orthologs of 54 candidate risk genes within these loci

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service