Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200797

Sigma-Aldrich

Anti-HSP70 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Heat shock 70 kDa protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~70 kDa

species reactivity

chicken, rat, canine, human, monkey, mouse, bovine

technique(s)

immunoblotting: 1:250-1:500 using different cell extracts
immunoprecipitation (IP): 10-20 μg/test

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA1A(3303)

General description

Heat Shock Proteins 70 (HSP70), also known as HSPA1A (heat shock protein family A (Hsp70) member 1A) or HSPA1B (heat shock protein family A (Hsp70) member 1B) is encoded by the gene mapped to human chromosome 6p21.3. xHSP70 is a highly conserved protein family with four members such as the constitutive (or cognate) HSP73, the stress-inducible HSP72, the glucose regulated proteins (grp)78 and grp75. During heat shock, HSP70s concentrate in the cell nuclei and return to the cytoplasm when the shock is removed.

Immunogen

Synthetic peptide from the internal region of human HSP70 protein, conjugated to KLH

Application

The antibody has been used in various immunochemical techniques including immunoblotting (∼70 kDa) and immunoprecipitation.Detection of the HSP70 band by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

Heat Shock Proteins 70 (HSP70) plays a key role in stress response, including heat stress, in every organism in which the proteins have been examined. These proteins also facilitates a wide range of folding processes, including the folding and assembly of newly synthesized proteins, refolding of misfolded and aggregated proteins, membrane translocation of organellar and secretory proteins, and control of the activity of regulatory proteins. In addition, HSP70 also play a cytoprotective role in neurodegenerative diseases such as Alzheimer′s, Parkinson′s, Huntington′s, ALS and others. Unregulated expression of HSP70s leads to the development of several types of cancer, such as malignant melanoma and renal cell cancer.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georgios D Lianos et al.
Cancer letters, 360(2), 114-118 (2015-02-28)
Heat shock proteins (HSPs) are an evolutionary family of proteins that act as molecular chaperones. According to their size they have been classified into the following families; HSP90, HSP70, HSP60, HSP40 and HSP27. They prevent the formation of nonspecific protein
Hsp70 dynamics in vivo: effect of heat shock and protein aggregation
Zeng X, et al.
Journal of Cell Science, 117(21), 4991-5000 (2004)
M Tavaria et al.
Cell stress & chaperones, 1(1), 23-28 (1996-04-01)
The human Hsp70 family encompasses at least 11 genes which encode a group of highly related proteins. These proteins include both cognate and highly inducible members, at least some of which act as molecular chaperones. The location of cognate Hsp70s
Heat shock protein 70 in Alzheimer?s disease
Lu RC, et al.
BioMed Research International, 2014(21) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service