Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200711

Sigma-Aldrich

Anti-Human Serum Albumin antibody, Mouse monoclonal

clone HSA-11, purified from hybridoma cell culture

Synonym(s):

ALB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HSA-11, monoclonal

form

buffered aqueous solution

mol wt

~70 kDa

species reactivity

gibbon, monkey, baboon

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 2.5-5 ng/mL using human serum
immunohistochemistry: suitable
indirect ELISA: 0.2-0.4 μg/mL using 10 μg/ml Human Serum Albumin for coating

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALB(213)

General description

Anti-human serum albumin antibody, mouse monoclonal (mouse IgG2a isotype) is derived from the HSA-11 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mouse immunized with human serum albumin (HSA). Monoclonal anti-human serum albumin specifically recognizes human albumin derived from sera, plasma or liver cells.
HSA is encoded by the gene mapped to human chromosome 4q13.3. It is characterized with three homologous domains that assemble to form a heart-shaped molecule. HSA constitutes around 60-65% of total serum protein.
The human serum albumin (HSA) gene is located on human chromosome 4. It encodes a globular protein that is the most prominent protein in human sera. It reaches a total concentration of about 60% of the total proteins in the blood serum. The globular protein is composed of 585 amino acids and comprises of three globular domains that resemble each other in structure, each containing two subdomains.

Specificity

Anti-Human Serum Albumin antibody recognizes human albumin derived from sera, plasma or liver cells.

Immunogen

Human Serum Albumin

Application

The antibody can be used in enzyme linked immunosorbent assay (ELISA), immunoblotting (~70 kDa) and immunohistochemistry.

Biochem/physiol Actions

Human serum albumin (HAS) serves as a transport and depot protein for several endogenous ligands including fatty acids, hormones, steroids, bilirubin and heme. High intravascular concentration of HSA enables it to play an essential role in exerting colloid osmotic pressure and thereby to regulate the oncotic pressure and volume of blood. It also acts as an antioxidant and is required for exerting Donnan effect in the capillaries. Mutations in the gene is associated with the development of bisalbuminaemia or alloalbuminaemia. Deficiency of the gene leads to analbuminaemia.
Human serum albumin (HSA) is a single non-glycosylated chain that has excellent binding capacity for various endogenous and exogenous ligands. It functions as a plasma transporter molecule and primarily binds to non-esterified long-chain fatty acids. It also binds to and transports various metabolites, such as bilirubin, steroid hormones, thyroxine, tryptophan, certain vitamins and metal ions within the body. It also has the ability to bind to several drugs and affects their pharmacokinetics and pharmacodynamics. HSA functions as a NO-carrier and is also responsible for the antioxidant capacity of human serum. In cases of acute hemolysis, HSA binds to heme in the blood stream and transports to hemopexin, where in it is reabsorbed by parenchymal liver cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congenital analbuminaemia: Molecular defects and biochemical and clinical aspects
Minchiotti L, et al.
Biochimica et Biophysica Acta, 1830, 5494-5502 (2013)
Aldo Prawira et al.
Liver international : official journal of the International Association for the Study of the Liver, 41(3), 608-620 (2020-11-13)
Infigratinib is a pan-FGFR (fibroblast growth factor receptor) inhibitor that has shown encouraging activity in FGFR-dependent hepatocellular carcinoma (HCC) models. However, long-term treatment results in the emergence of resistant colonies. We sought to understand the mechanisms behind infigratinib-induced tumour cell
G Chevalier et al.
The American journal of pathology, 152(6), 1563-1575 (1998-06-17)
We previously established that the expression of the human nov gene (novH) was altered in Wilms' tumors and that levels of novH and WT1 mRNA were inversely correlated in individual Wilms' tumors. Insofar as novH has been shown to be
Beyond expansion: structural studies on the transport roles of human serum albumin.
Curry S.
Vox Sanguinis, 83(s1), 315-319 (2002)
Atomic structure and chemistry of human serum albumin
He XM and Carter DC
Nature, 358, 209-215 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service