Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200450

Sigma-Aldrich

Anti-SOX11 (C-terminal) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-SRY (sex determining region Y)-box 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human

technique(s)

immunohistochemistry: 1:100-1:200 using formalin-fixed paraffin-embedded rat brain and cerebellum.
indirect immunofluorescence: 1:100-1:200 using human HeLa cells.
western blot: 1:4000-1:8000 using whole extracts of HEK-293T cells overexpressing human recombinant SOX11

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX11(6664)

General description

Sex determining region Y (SRY)-box 11 (SOX11) protein belongs to the SRY-related high mobility group (HMG)-box (SOX) family of transcription factors. This protein is localized in adult immature neurons but is normally not expressed in any other adult tissue. SOX11 gene is located on the human chromosome 2p25.2.

Specificity

Anti-SOX11 (C-terminal) recognizes human and rat SOX11.

Immunogen

peptide corresponding to the C-terminal region of human SOX11, conjugated to KLH. The corresponding sequence is identical in monkey, bovine and pig and differs by a single amino acid in mouse and rat.

Application

Anti-SOX11 (C-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunofluorescence
  • immunohistochemistry

Biochem/physiol Actions

Sex determining region Y (SRY)-box 11 (SOX11) plays a role in the regulation of tissue remodeling during embryogenesis which is essential for neurogenesis. This protein is also involved in tumorigenesis. SOX11 protein also plays a role in the determination of cell fate. Upregulation of the SOX11 gene leads to ovarian cancer, brain tumor, medulloblastomas and gliomas. This protein functions as a biomarker in mantle cell lymphoma (MCL), acute lymphoblastic leukemia (ALL) and Burkitt lymphomas (BL).

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Palomino Lago et al.
Animals : an open access journal from MDPI, 14(1) (2024-01-11)
Fractures caused by bone overloading are a leading cause of euthanasia in Thoroughbred racehorses. The risk of fatal fracture has been shown to be influenced by both environmental and genetic factors but, to date, no specific genetic mechanisms underpinning fractures
Maria Carmela Vegliante et al.
PloS one, 6(6), e21382-e21382 (2011-07-09)
Recent studies have shown aberrant expression of SOX11 in various types of aggressive B-cell neoplasms. To elucidate the molecular mechanisms leading to such deregulation, we performed a comprehensive SOX11 gene expression and epigenetic study in stem cells, normal hematopoietic cells
Xiao Wang et al.
PloS one, 5(11), e14085-e14085 (2010-12-03)
The SRY (sex determining region Y)-box 11 (SOX11) gene, located on chromosome 2p25, encodes for a transcription factor that is involved in tissue remodeling during embryogenesis and is crucial for neurogenesis. The role for SOX11 in hematopoiesis has not yet
Lakshmi Pillai-Kastoori et al.
PLoS genetics, 10(7), e1004491-e1004491 (2014-07-11)
Ocular coloboma is a sight-threatening malformation caused by failure of the choroid fissure to close during morphogenesis of the eye, and is frequently associated with additional anomalies, including microphthalmia and cataracts. Although Hedgehog signaling is known to play a critical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service