Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900334

Sigma-Aldrich

Anti-HSP90AA1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

heat shock protein hsp 90-alpha, hsp 86, hsp90a, hspc1, hspca

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse, chicken, monkey, Drosophila

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSP90AA1(3320)

General description

Heat shock protein 90 (Hsp90α) gene is mapped to human chromosome 14q32.31. Hsp90α is an isoform of Hsp90 and is a cytoplasmic protein that comprises three domains, namely C-terminal, middle domain, and N-terminal nucleotide-binding domain. This protein is associated with membrane microdomains and lipid rafts.

Immunogen

Synthetic peptide from human HSP90AA1 / Hsp90.

Application

Anti-HSP90AA1 antibody produced in rabbit has been used in immunohistochemistry at a dilution of 1:400.

Biochem/physiol Actions

Heat shock protein 90 alpha family class A member 1 (HSP90AA1 or Hsp90α) is upregulated during stress and helps in survival from membrane stress, especially during drug targeting. It elicits cytoprotective function and mediates protein homeostasis in the cytoplasm. An elevated level of Hsp90α is observed during chronic obstructive pulmonary disease (COPD). Hsp90α also favors spermatocyte maturation and human ether-a-go-go-related gene (hERG) ion channel maturation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.02 M potassium phosphate, 0.15 M sodium chloride, pH 7.2, 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiichi Koshizuka et al.
Auris, nasus, larynx, 45(4), 854-865 (2017-12-14)
Our recent studies have revealed that both strands of pre-miRNAs, the guide strand and the passenger strand, are involved in cancer pathogenesis. Analyses of miRNA expression signatures by RNA sequencing in head and neck squamous cell carcinoma (HNSCC) showed that
Abbey D Zuehlke et al.
Gene, 570(1), 8-16 (2015-06-14)
Heat shock protein 90α (Hsp90α), encoded by the HSP90AA1 gene, is the stress inducible isoform of the molecular chaperone Hsp90. Hsp90α is regulated differently and has different functions when compared to the constitutively expressed Hsp90β isoform, despite high amino acid
Pengpeng Li et al.
Biochimica et biophysica acta. Biomembranes, 1861(2), 457-465 (2018-12-06)
During heat shock conditions, structural changes in cellular membranes may lead to cell death. Hsp90AA1 and other heat shock proteins involved in membranes are responsible for protecting membrane stabilization. However, the membrane binding mechanism of Hsp90AA1 remains largely uncharacterized. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service