Skip to Content
Merck
All Photos(7)

Key Documents

SAB2702052

Sigma-Aldrich

Anti-SMAD3 antibody produced in rabbit

Synonym(s):

JV15-2, LDS1C, LDS3, MADH3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

48 kDa

species reactivity

human, mouse, rat

concentration

0.78 mg/mL

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: 1000-10000

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SMAD3(4088)

Immunogen

Recombinant fragment corresponding to a region within amino acids 122 and 395 of SMAD3 (Uniprot ID#P84022)

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, IP: 1:100-1:500, WB: 1:1000-1:10000, ChIP assay: Assay-dependent dilution. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene ′mothers against decapentaplegic′ (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soo Youn Cho et al.
Medical oncology (Northwood, London, England), 31(11), 236-236 (2014-10-01)
Smad3 functions as an integrator of diverse signaling, including transforming growth factor β signaling and the function of Smad3 is complexly regulated by differential phosphorylation at various sites of Smad3. Despite the importance of Smad3 and its various phosphoisoforms, their
Yunchuan Li et al.
Chinese medical journal, 127(19), 3449-3453 (2014-10-02)
Aberrant epithelial repair has been observed in chronic rhinosinusitis (CRS) patients; however, the mechanism of epithelial cell repair regulation is unclear. Epidermal growth factor (EGF) plays an important role in regulating epithelial cell repair in lower airway and may be
Nicolás Tobar et al.
BMC cancer, 14, 640-640 (2014-09-02)
Hard consistency, developed under the influence of tumor cell factors, is a characteristic feature of a breast tumor. Activation of resident fibroblasts leading to a myofibroblast phenotype is the principal feature that orchestrates this fibrotic process. The aim of this
Jinyi Han et al.
Experimental biology and medicine (Maywood, N.J.), 239(3), 272-283 (2014-02-07)
All-trans retinoic acid (ATRA) has been used for the treatment of acute promyelocytic leukemia. It remains unclear, however, whether ATRA affects cyclooxygenase-2 (COX-2; an enzyme involved in prostaglandin production), PGE2, and thromboxane A2 (TXA2) (metabolic products of COX-2) by a
Peng Li et al.
Stem cell research & therapy, 10(1), 144-144 (2019-05-23)
Non-small cell lung cancer (NSCLC) is the second most prevalent cause of cancer-related fatality. Long non-coding RNAs (lncRNAs) have been observed to exercise functions in NSCLC. Here, the current study aimed to explore the potential mechanism of lncRNA MBNL1-AS1 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service