Skip to Content
Merck
All Photos(2)

Key Documents

SAB2500845

Sigma-Aldrich

Anti-Pyruvate Carboxylase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PCB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat, canine, pig, bovine

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PC(5091)

General description

Pyruvate carboxylase (PC), a mitochondrial enzyme, is a nuclear-encoded protein. Expression of biologically active PC is seen in skeletal muscle. PC is a homotetramer in vertebrates, with each subunit having biotin carboxylase (BC), carboxyltransferase (CT), and biotin-carboxyl carrier protein (BCCP) domains and a covalently linked biotin cofactor. The PC gene is located on human chromosome 11q13.2.

Immunogen

Peptide with sequence C-KFKEVKKAYVEANQ from the internal region of the protein sequence according to NP_000911.2; NP_001035806.1; NP_071504.2.

Application

Anti-Pyruvate Carboxylase antibody produced in goat has been used in immunoblotting.

Biochem/physiol Actions

Pyruvate carboxylase (PC) enzyme converts pyruvate to oxaloacetate. It plays an important role in mitochondrial metabolism in all cells and its absence is lethal. Expression of PC is highly essential for the development of pulmonary tumors. PC is involved in the pulmonary tropism of metastatic breast cancer. It is associated with the pathogenesis of ovarian cancer. Lower expression of PC can block cell proliferation, progression of the cell cycle, migration, and invasion of cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongkai Shang et al.
Aging, 12(21), 21874-21889 (2020-11-13)
The aim of this study was to explore prognosis-related biomarkers and underlying mechanisms during ovarian carcinoma progression and development. mRNA expression profiles and GSE49997 dataset were downloaded. Survival analyses were performed for genes with high expression levels. Expression level of
Ariane D Minet et al.
Biochemical and biophysical research communications, 402(2), 196-197 (2010-09-03)
Pyruvate carboxylase (PC) is a mitochondrial enzyme that catalyses the carboxylation of pyruvate to oxaloacetate thereby allowing supplementation of citric acid cycle intermediates. The presence of PC in skeletal muscle is controversial. We report here, that PC protein is easily
Arun Nandwani et al.
Redox biology, 69, 102989-102989 (2023-12-16)
Inspite of exerting independent cellular functions, the endoplasmic-reticulum (ER) and the mitochondria also physically connect at specific sites termed mitochondria-associated ER membranes (MAMs) and these sites consist of several tethering proteins that play varied roles in diverse cellular processes. However
Aparna Shinde et al.
Breast cancer research : BCR, 20(1), 76-76 (2018-07-15)
Overcoming systemic dormancy and initiating secondary tumor grow under unique microenvironmental conditions is a major rate-limiting step in metastatic progression. Disseminated tumor cells encounter major changes in nutrient supplies and oxidative stresses compared to the primary tumor and must demonstrate
Raju Ahmed et al.
Journal of immunological methods, 406, 143-147 (2014-03-25)
Endogenous biotin or biotinylated protein binding activity is a major drawback to biotin-avidin/streptavidin detection system. The avidin/streptavidin conjugate used to detect the complex of the biotinylated secondary antibody and the primary antibody binds to endogenous biotin or biotinylated proteins leading

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service