Skip to Content
Merck
All Photos(6)

Key Documents

SAB2108668

Sigma-Aldrich

Anti-GAPDH

affinity isolated antibody

Synonym(s):

Anti- GAPD, Anti- MGC88685, Anti-G3PD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

rat, goat, guinea pig, human, rabbit, horse, sheep, dog

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_002046

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)

Immunogen

Synthetic peptide directed towards the N terminal region of human GAPDH

Biochem/physiol Actions

GAPDH catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. Many pseudogenes similar to this locus are present in the human genome.Glyceraldehyde-3-phosphate dehydrogenase catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. A GAPD pseudogene has been mapped to Xp21-p11 and 15 GAPD-like loci have been identified. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: IDLNYMVYMFQYDSTHGKFHGTVKAENGKLVINGNPITIFQERDPSKIKW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Qu et al.
Reproductive sciences (Thousand Oaks, Calif.), 27(6), 1276-1284 (2020-02-13)
In response to cytosolic DNA, stimulator of interferon gene (STING) initiates and orchestrates host's innate immunity by inducing type I interferon. Since endometriosis is a chronic inflammatory disorder, we sought to determine whether STING pathway is activated in ectopic endometrium
Luca Ambrosio et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 33(5), 1713-1727 (2024-02-28)
To investigate the therapeutic potential of extracellular vesicles (EVs) derived from human nucleus pulposus cells (NPCs), with a specific emphasis on Tie2-enhanced NPCs, compared to EVs derived from human bone marrow-derived mesenchymal stromal cells (BM-MSCs) in a coccygeal intervertebral disc
Christelle Damon-Soubeyrand et al.
eLife, 12 (2023-06-13)
Long considered an accessory tubule of the male reproductive system, the epididymis is proving to be a key determinant of male fertility. In addition to its secretory role in ensuring functional maturation and survival of spermatozoa, the epididymis has a
Baijun Li et al.
Oncology letters, 22(6), 851-851 (2021-11-05)
Procollagen-lysine, 2-oxoglutarate 5-dioxygenase 1 (PLOD1) is an enzyme that catalyzes the hydroxylation of lysyl residues in collagen-like peptides, and is responsible for the stability of intermolecular crosslinks. High PLOD1 mRNA levels have been determined to be prognostically significant in numerous
Qiang Xiao et al.
Molecular medicine reports, 19(3), 2377-2385 (2019-01-22)
Leukemia inhibitory factor (LIF) is a multifunctional cytokine. The present study aimed to determine the expression and effects of LIF on nucleus pulposus generation. Degenerated nucleus pulposus samples were obtained from animal models and patients with lumbar intervertebral disc herniation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service