Skip to Content
Merck
All Photos(8)

Key Documents

SAB2108004

Sigma-Aldrich

Anti-BDNF antibody produced in rabbit

Synonym(s):

Anti-ANON2, Anti-BULN2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

27kDa

species reactivity

horse, rat, rabbit, dog, mouse, human, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

General description

The brain derived neurotrophic factor (BDNF) gene is mapped to human chromosome 11p14.1. BDNF is a member of the neurotrophin family of growth factors. The gene encodes a precursor protein, proBDNF. Mature BDNF (mBDNF) is synthesized by post-translational cleavage of proBDNF. Both proBDNF and mBDNF play crucial roles in cellular signaling.

Immunogen

Synthetic peptide directed towards the middle region of human BDNF

Application

Anti-BDNF antibody produced in rabbit has been used in immunohistochemistry and western blotting.

Biochem/physiol Actions

Brain derived neurotrophic factor (BDNF) is involved in hippocampal function and verbal episodic memory in humans. Hence, variation in the BDNF gene expression alters these neurological functions. BDNF also plays a vital role in vascular function and participates in angiogenesis via the specific receptor tropomyosin-related kinase B (TrkB). It is involved in the pathogenesis of Alzheimer′s disease. ProBDNF interacts with p75 neurotrophin receptor, leading to long-term depression in the hippocampus.

Sequence

Synthetic peptide located within the following region: EWVTAADKKTAVDMSGGTVTVLEKVPVSKGQLKQYFYETKCNPMGYTKEG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of nerve growth factor and brain-derived neurotrophic factor in astrocytomas.
Liu TT, et al.
Oncology Letters, 15, 533-537 (2018)
Running wheel exercise reduces a-synuclein aggregation and improves motor and cognitive function in a transgenic mouse model of Parkinson's disease
Zhou W, et al.
PLoS ONE, 12 (2017)
Daniela Rodrigues-Amorim et al.
Frontiers in psychiatry, 10, 885-885 (2019-12-19)
Schizophrenia is a severe and disabling psychiatric disorder with a complex and multifactorial etiology. The lack of consensus regarding the multifaceted dysfunction of this ailment has increased the need to explore new research lines. This research makes use of proteomics
Kíssila Rabelo et al.
Frontiers in immunology, 11, 2146-2146 (2020-09-29)
In Brazil, an epidemic of Zika virus (ZIKV) infections was declared in 2015 that coincided with alarming reports of microcephaly in newborns associated with mother infection. Although the virus has placental tropism, changes in the tissue morphology and immunity of
Bin Liu et al.
Journal of ovarian research, 16(1), 83-83 (2023-04-28)
Brain-derived neurotrophic factor (BDNF) plays an important role in ovarian function including follicle development and oocyte maturation, and embryonic development. However, whether BDNF treatment can reimpose ovarian aging and impaired fertility remains elusive. In this study, we investigated the reproductive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service