Skip to Content
Merck
All Photos(4)

Key Documents

SAB2104243

Sigma-Aldrich

Anti-TRPV4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-OTRPC4, Anti-TRP12, Anti-VR-OAC, Anti-VRL2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

91 kDa

species reactivity

human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPV4(59341)

Immunogen

Synthetic peptide directed towards the middle region of human TRPV4

Biochem/physiol Actions

TRPV4 is a member of the OSM9-like transient receptor potential channel (OTRPC) subfamily in the transient receptor potential (TRP) superfamily of ion channels. TRPV4 is a Ca2+-permeable, nonselective cation channel that is thought to be involved in the regulation of systemic osmotic pressure.This gene encodes a member of the OSM9-like transient receptor potential channel (OTRPC) subfamily in the transient receptor potential (TRP) superfamily of ion channels. The encoded protein is a Ca2+-permeable, nonselective cation channel that is thought to be involved in the regulation of systemic osmotic pressure. Two transcript variants encoding different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: RVDEVNWSHWNQNLGIINEDPGKNETYQYYGFSHTVGRLRRDRWSSVVPR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Shi et al.
Pharmaceutical biology, 61(1), 1000-1012 (2023-07-06)
Hyperoside (Hyp), one of the active flavones from Rhododendron (Ericaceae), has beneficial effects against cerebrovascular disease. However, the effect of Hyp on vasodilatation has not been elucidated. To explore the effect of Hyp on vasodilatation in the cerebral basilar artery
Kathia Beddek et al.
Physiological reports, 9(3), e14613-e14613 (2021-01-30)
The Transient Receptor Potential Vanilloid 4 (TRPV4) of endothelial cells contributes to many important functions including the regulation of Ca2+ homeostasis, cell volume, endothelial barrier permeability, and smooth muscle tone. However, its role in the transition of endothelial cells toward
Barbara Barile et al.
Frontiers in cellular neuroscience, 17, 1247761-1247761 (2023-09-18)
Despite of the major role of aquaporin (AQP) water channels in controlling transmembrane water fluxes, alternative ways for modulating water permeation have been proposed. In the Central Nervous System (CNS), Aquaporin-4 (AQP4) is reported to be functionally coupled with the
Yan Zhang et al.
American journal of physiology. Gastrointestinal and liver physiology, 303(12), G1365-G1372 (2012-10-23)
Transient receptor potential vanilloid subtype 4 (TRPV4) is a ligand-gated nonselective cation channel that participates in the transduction of mechanical and osmotic stimuli in different tissues. TRPV4 is activated by endogenous arachidonic acid metabolites, 4α-phorbol-12,13 didecanoate, GSK1016790A, moderate heat, and
S Fu et al.
Osteoarthritis and cartilage, 29(1), 89-99 (2021-01-05)
Cartilage health is maintained in response to a range of mechanical stimuli including compressive, shear and tensile strains and associated alterations in osmolality. The osmotic-sensitive ion channel Transient Receptor Potential Vanilloid 4 (TRPV4) is required for mechanotransduction. Mechanical stimuli inhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service