Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102330

Sigma-Aldrich

Anti-STXBP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HUNC18, Anti-MUNC18-1, Anti-RbSec1, Anti-Syntaxin binding protein 1, Anti-UNC18

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

67 kDa

species reactivity

human, horse, dog, bovine, rabbit, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STXBP1(6812)

General description

The gene STXBP1 (syntaxin binding protein 1) is mapped to human chromosome 9q34.1. The encoded protein is 594 amino acids long, which is expressed in all tissues with highest levels in retina and cerebellum.

Immunogen

Synthetic peptide directed towards the middle region of human STXBP1

Biochem/physiol Actions

The gene STXBP1 (syntaxin binding protein 1) encodes a regulatory protein of the components of the SNARE (soluble NSF/ N-ethylmaleimide-sensitive factor attachment protein receptor) complex that are crucial for neurotransmission. It is a neuronal protein that functions in synaptic vesicle release. Mutations in this gene have been associated with early infantile epileptic encephalopathy. It causes autosomal dominant mental retardation and nonsyndromic epilepsy.

Sequence

Synthetic peptide located within the following region: TRSSASFSTTAVSARYGHWHKNKAPGEYRSGPRLIIFILGGVSLNEMRCA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xhuliana Kajana et al.
Biomolecules, 13(12) (2023-12-23)
Cerebrospinal fluid (CSF) is a biochemical-clinical window into the brain. Unfortunately, its wide dynamic range, low protein concentration, and small sample quantity significantly limit the possibility of using it routinely. Extraventricular drainage (EVD) of CSF allows us to solve quantitative
D A Swanson et al.
Genomics, 48(3), 373-376 (1998-04-18)
In a screen designed to identify genes expressed preferentially in retina, we identified a cDNA encoding the human ortholog of rat STXBP1 (n-Sec1, Munc-18-1, rbSec1), a protein implicated in vesicle trafficking and neurotransmitter release. This protein also has similarity to
Hirotomo Saitsu et al.
Nature genetics, 40(6), 782-788 (2008-05-13)
Early infantile epileptic encephalopathy with suppression-burst (EIEE), also known as Ohtahara syndrome, is one of the most severe and earliest forms of epilepsy. Using array-based comparative genomic hybridization, we found a de novo 2.0-Mb microdeletion at 9q33.3-q34.11 in a girl
Fadi F Hamdan et al.
Annals of neurology, 65(6), 748-753 (2009-06-27)
We sequenced genes coding for components of the SNARE complex (STX1A, VAMP2, SNAP25) and their regulatory proteins (STXBP1/Munc18-1, SYT1), which are essential for neurotransmission, in 95 patients with idiopathic mental retardation. We identified de novo mutations in STXBP1 (nonsense, p.R388X;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service